Skip to Content
Merck
All Photos(3)

Key Documents

216623

Sigma-Aldrich

Sodium chromate tetrahydrate

99%

Synonym(s):

Disodium chromate tetrahydrate, Sodium chromate(VI) tetrahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2CrO4 · 4H2O
CAS Number:
Molecular Weight:
234.03
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22
Assay:
99%
form:
solid

Quality Level

Assay

99%

form

solid

SMILES string

O.O.O.O.[Na+].[Na+].[O-][Cr]([O-])(=O)=O

InChI

1S/Cr.2Na.4H2O.4O/h;;;4*1H2;;;;/q;2*+1;;;;;;;2*-1

InChI key

AVHUVTPOXZEIRG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Sodium chromate tetrahydrate can be used as an oxidant in organic synthesis and also as a carrier electrolyte in the capillary electrophoresis.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Muta. 1B - Repr. 1B - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A study of the pollution of the Aries River (Romania) using capillary electrophoresis as analytical technique.
Forray F L and Hallbauer D K
Environmental Geology, 39(12), 1372-1384 (2000)
Sodium chromate oxidation of enol benzoates. Allylic oxidation versus a novel rearrangement.
Goldblum A and Mechoulam R
Journal of the Chemical Society. Perkin Transactions 1, (17), 1889-1898 (1977)
Judith M Myers et al.
Toxicology, 246(2-3), 222-233 (2008-03-11)
Hexavalent chromium [Cr(VI)] species such as chromates are cytotoxic. Inhalational exposure is a primary concern in many Cr-related industries and their immediate environments, and bronchial epithelial cells are directly exposed to inhaled Cr(VI). Chromates are readily taken up by cells
Seung-Hee Maeng et al.
Mutation research, 539(1-2), 109-116 (2003-09-02)
According to the toxicological and epidemiological studies, hexavalent chromium (Cr) is associated with increase of lung cancer risk. Genotoxic effects, such as chromosomal aberrations, and cellular oxidative DNA damage by reactive oxygen species produced by hexavalent Cr exposure may play
Versatility of capillary electrophoresis of anions with a high mobility chromate electrolyte.
Oehrle S A.
Journal of Chromatography A, 671(1-2), 383-387 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service