144509
N-Phenylanthranilic acid
98%
Synonym(s):
2-(Phenylamino)benzoic acid, Diphenylamine-2-carboxylic acid, DpC
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
2-(C6H5NH)C6H4CO2H
CAS Number:
Molecular Weight:
213.23
Beilstein:
1456607
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
form
powder
reaction suitability
reaction type: solution phase peptide synthesis
mp
182-185 °C (lit.)
application(s)
peptide synthesis
SMILES string
OC(=O)c1ccccc1Nc2ccccc2
InChI
1S/C13H11NO2/c15-13(16)11-8-4-5-9-12(11)14-10-6-2-1-3-7-10/h1-9,14H,(H,15,16)
InChI key
ZWJINEZUASEZBH-UHFFFAOYSA-N
Gene Information
human ... TTR(7276)
Looking for similar products? Visit Product Comparison Guide
Related Categories
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Jana Obajdin et al.
Toxicological sciences : an official journal of the Society of Toxicology, 166(2), 441-450 (2018-09-15)
Identification of improved translatable biomarkers of nephrotoxicity is an unmet safety biomarker need. Fatty-acid-binding protein 4 (FABP4) was previously found to be associated with clinical renal dysfunction and was proposed as a biomarker of glomerular damage. The aim of this
Walter E Finkbeiner et al.
American journal of physiology. Lung cellular and molecular physiology, 301(4), L402-L414 (2011-07-05)
We investigated how cystic fibrosis (CF) alters the relationship between Cl(-) and mucin secretion in cultures of non-CF and CF human tracheobronchial gland mucous (HTGM and CFTGM, respectively) cells. Biochemical studies showed that HTMG cells secreted typical airway mucins, and
Asher Peretz et al.
Proceedings of the National Academy of Sciences of the United States of America, 107(35), 15637-15642 (2010-08-18)
The pore and gate regions of voltage-gated cation channels have been often targeted with drugs acting as channel modulators. In contrast, the voltage-sensing domain (VSD) was practically not exploited for therapeutic purposes, although it is the target of various toxins.
Ernie Harpur et al.
Toxicological sciences : an official journal of the Society of Toxicology, 122(2), 235-252 (2011-05-20)
This study reports the evaluation of four urinary biomarkers of renal toxicity, α-glutathione-S-transferase (α-GST), μ-GST, clusterin, and renal papillary antigen-1 (RPA-1), in male Sprague-Dawley and Han-Wistar rats given cisplatin, gentamicin, or N-phenylanthranilic acid (NPAA). Kidney injury was diagnosed histopathologically, according
Olga J Baker et al.
Tissue engineering. Part C, Methods, 16(5), 1135-1144 (2010-02-04)
The use of polarized salivary gland cell monolayers has contributed to our understanding of salivary gland physiology. However, these cell models are not representative of glandular epithelium in vivo, and, therefore, are not ideal for investigating salivary epithelial functions. The
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service