Skip to Content
Merck
All Photos(2)

Key Documents

206555

Sigma-Aldrich

4-Iodotoluene

99%

Synonym(s):

1-Iodo-4-methylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4I
CAS Number:
Molecular Weight:
218.03
Beilstein:
1903637
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

99%

form

solid

bp

211.5 °C (lit.)

mp

33-35 °C (lit.)

functional group

iodo

SMILES string

Cc1ccc(I)cc1

InChI

1S/C7H7I/c1-6-2-4-7(8)5-3-6/h2-5H,1H3

InChI key

UDHAWRUAECEBHC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

4-Iodotoluene undergoes Suzuki-Miyaura coupling reaction with phenylboronic acid catalyzed by (Ni,Mg)3Si2O5(OH)4 solid-solution nanotubes loaded with palladium[1]. Cobalt-catalyzed coupling of 4-iodotoluene with thiophenols and alkanethiols has been investigated[2]. Palladium/copper-catalyzed Sonogashira cross-coupling reaction of 4-iodotoluene with phenylacetylene has been studied[3].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

194.0 °F - closed cup

Flash Point(C)

90 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wancheng Zhu et al.
Inorganic chemistry, 51(11), 6020-6031 (2012-05-16)
(Ni(1-x),Mg(x))(3)Si(2)O(5)(OH)(4) solid-solution nanotubes (NTs) with tunable compositions were hydrothermally synthesized by altering the molar ratio of Mg(2+) to Ni(2+). The as-synthesized NTs were loaded with sub-0.06 wt % palladium (Pd; ∼0.045 wt %) for Suzuki-Miyaura (SM) coupling reactions between iodobenzene
Takashi Mino et al.
The Journal of organic chemistry, 71(25), 9499-9502 (2006-12-02)
Palladium/copper-catalyzed Sonogashira cross-coupling reaction of aryl halides with a variety of terminal alkynes under amine-free conditions in dimethylformamide (DMF) at 80 degrees C gave internal arylated alkynes using PdCl2(MeCN)2 with phosphine-free hydrazone 2a as a ligand and CuI as the
Ying-Chieh Wong et al.
Organic letters, 8(24), 5613-5616 (2006-11-17)
A new cobalt-catalyzed coupling of aryl halides with thiophenols and alkanethiols is reported. A variety of aryl sulfides can be prepared in excellent yields under mild reaction conditions using 1-2 mol % of CoI2(dppe) and Zn. This new cobalt-catalyzed coupling
V Kolaříková et al.
Dalton transactions (Cambridge, England : 2003), 44(45), 19663-19673 (2015-09-17)
Using three different approaches, racemic 1-(perfluoroalkyl)ethylamines were synthesized from perfluoroalkyl iodides or perfluoroalkanoic acids, and further transformed to the corresponding N,N'-disubstituted ethane-1,2-diimines and ethane-1,2-diamines as mixtures of diastereoisomers. Their cyclization afforded imidazolium or dihydroimidazolium salts, which led to silver or
Zengyan Wei et al.
Nature communications, 5, 3870-3870 (2014-05-16)
The shape-controlled synthesis of nanoparticles was established in single-phase solutions by controlling growth directions of crystalline facets on seed nanocrystals kinetically; however, it was difficult to rationally predict and design nanoparticle shapes. Here we introduce a methodology to fabricate nanoparticles

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service