Skip to Content
Merck
All Photos(3)

Documents

111929

Sigma-Aldrich

4-Chlorotoluene

98%

Synonym(s):

1-Chloro-4-methylbenzene, 1-Methyl-4-chlorobenzene, 4-Methyl-1-chlorobenzene, 4-Methylchlorobenzene, 4-Methylphenyl chloride, p-Chloromethylbenzene, p-Chlorotoluene, p-Methylchlorobenzene, p-Tolyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4Cl
CAS Number:
Molecular Weight:
126.58
Beilstein:
1903635
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39050207
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.38 (vs air)

vapor pressure

10 mmHg ( 45 °C)

Assay

98%

form

liquid

impurities

≤2% 2-chlorotoluene

refractive index

n20/D 1.52 (lit.)

bp

162 °C (lit.)

mp

6-8 °C (lit.)

density

1.07 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(Cl)cc1

InChI

1S/C7H7Cl/c1-6-2-4-7(8)5-3-6/h2-5H,1H3

InChI key

NPDACUSDTOMAMK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The reaction of 4-chlorotouleneand phenylboronic acid with Pd(OAc)2/L•2HX as the catalyst is the model reaction for determining the efficiencies of bisimidazolylidene (L) ligand precursors.

Application

The product was used for the preparation model gas of 4-chlorotoulene by passing synthesized air into liquid 4-chlorotoulene.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Flam. Liq. 3 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

123.8 °F - closed cup

Flash Point(C)

51 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Palladium-bisimidazol-2-ylidene complexes as catalysts for general and efficient Suzuki cross-coupling reactions of aryl chlorides with arylboronic acids.
Zhang C and Trudell ML.
Tetrahedron Letters, 41(5), 595-598 (2000)
Organic pollutants treatment in gas phase by using electron beam generated non-thermal plasma reactor.
Sun, Y, et al.
Chem. Listy, 102(16), s1524-s1528 (2008)
T Nishio et al.
Applied microbiology and biotechnology, 55(3), 321-325 (2001-05-09)
p-Cymene monooxygenase (CMO) from Pseudomonas putida F1 consists of a hydroxylase (CymA1) and a reductase component (CymA2) which initiate pcymene (p-isopropyltoluene) catabolism by oxidation of the methyl group to p-isopropylbenzyl alcohol (p-cumic alcohol). To study the possible diverse range of
C Arunagiri et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 79(5), 1747-1756 (2011-06-18)
The FT-IR and FT-Raman spectra of 2-bromo-4-chlorotoluene (2B4CT) molecule have been recorded in the region 4000-400 cm(-1) and 3500-50 cm(-1), respectively. Optimized geometrical structures, harmonic vibrational frequencies, intensities, reduced mass, force constants and depolarization ratio have been computed by the
Ya-Jun Liu et al.
Physical chemistry chemical physics : PCCP, 7(23), 3938-3942 (2005-12-07)
Equilibrium geometries and vibrational frequencies of the ground and some excited states of p-chlorotoluene were calculated by the complete active space self-consistent field (CASSCF) method. Multi-reference CASSCF second order perturbation theory (MSCASPT2) calculations were performed on the vertical excitation energies

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service