Skip to Content
Merck
All Photos(1)

Documents

I7632

Sigma-Aldrich

Iodobenzene

98%

Synonym(s):

1-Iodobenzene, 4-Iodobenzene, Benzene iodide, Phenyl iodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5I
CAS Number:
Molecular Weight:
204.01
Beilstein:
1446140
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.62 (lit.)

bp

188 °C (lit.)

mp

−29 °C (lit.)

density

1.823 g/mL at 25 °C (lit.)

SMILES string

Ic1ccccc1

InChI

1S/C6H5I/c7-6-4-2-1-3-5-6/h1-5H

InChI key

SNHMUERNLJLMHN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Iodobenzene, also known as phenyl iodide, is commonly used as an arylating agent in various cross-coupling reactions.

Application

Iodobenzene is used in various C-C coupling reactions. It can be used in the preparation of iodobenzene dichloride, which is employed as an oxidant to oxidize alcohols to carbonyl compounds and as a chlorinating agent.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

170.6 °F - closed cup

Flash Point(C)

77 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heck reactions of iodobenzene and methyl acrylate with conventional supported palladium catalysts in the presence of organic and/and inorganic bases without ligands.
Zhao, Fengyu et al.
Chemistry?A European Journal , 6(5), 843-848 (2000)
Ring-Opening 1, 3-Dichlorination of Donor-Acceptor Cyclopropanes by Iodobenzene Dichloride.
Garve, Lennart KB et al.
Organic Letters, 16(21), 5804-5807 (2014)
Iodobenzene dichloride as a stoichiometric oxidant for the conversion of alcohols into carbonyl compounds; two facile methods for its preparation.
Zhao, Xue-Fei and Zhang, Chi
Synthesis, 2007(04), 551-557 (2007)
Size effects of PVP- Pd nanoparticles on the catalytic Suzuki reactions in aqueous solution.
Li, Yin et al.
Langmuir, 18(12), 4921-4925 (2002)
Catalysis by gold (I) and gold (III): a parallelism between homo-and heterogeneous catalysts for copper-free Sonogashira cross-coupling reactions.
Gonzalez-Arellano, Camino et al.
Angewandte Chemie (International Edition in English), 46(9), 1536-1538 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service