Skip to Content
Merck
All Photos(2)

Key Documents

238090

Sigma-Aldrich

1-Bromo-4-iodobenzene

98%

Synonym(s):

4-Bromo-1-iodobenzene, 4-Bromoiodobenzene, 4-Iodo-1-bromobenzene, 4-Iodophenyl bromide, p-Bromophenyl iodide, p-Iodobromobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4I
CAS Number:
Molecular Weight:
282.90
Beilstein:
1904545
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

bp

120-122 °C/14 mmHg (lit.)

mp

89-91 °C (lit.)

functional group

bromo

SMILES string

Brc1ccc(I)cc1

InChI

1S/C6H4BrI/c7-5-1-3-6(8)4-2-5/h1-4H

InChI key

UCCUXODGPMAHRL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Vapour pressure as a function of temperature for 1-bromo-4-iodobenzene has been studied using a diaphragm manometer and torsion mass-loss effusion.

Application

1-Bromo-4-iodobenzene has been employed:
  • as reagent for in situ desilylation and coupling of silylated alkynes
  • as starting reagent in the total syntheses of ent-conduramine A and ent-7-deoxypancratistatin (alkaloids)
  • as substrate in copper-free Sonogashira coupling in aqueous acetone
  • in synthesis of β,β,dibromostyrenes

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Representation and assessment of vapour pressure data; a novel approach applied to crystalline 1-bromo-4-chlorobenzene, 1-chloro-4-iodobenzene, and 1-bromo-4-iodobenzene.
Oonk HAJ, et al.
The Journal of Chemical Thermodynamics, 30(7), 897-907 (1998)
Synlett, 2061-2061 (2007)
Total syntheses of ent-conduramine A and ent -7-deoxypancratistatin.
Akgun H and Hudlicky T.
Tetrahedron Letters, 40(16), 3081-3084 (1999)
Synlett, 1843-1843 (2007)
Rapid bi-directional synthesis of oligo (1, 4-phenylene ethynylene) s.
Huang S and Tour JM.
Tetrahedron Letters, 40(17), 3447-3450 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service