Skip to Content
Merck
All Photos(4)

Key Documents

204854

Sigma-Aldrich

Vanadium(V) oxide

greener alternative

99.9% trace metals basis

Synonym(s):

Divanadium pentaoxide, Divanadium pentoxide, Pentaoxodivanadium, Vandia

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
V2O5
CAS Number:
Molecular Weight:
181.88
EC Number:
MDL number:
UNSPSC Code:
12352303
eCl@ss:
38180404
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

99.9% trace metals basis

form

powder

composition

V2O5

reaction suitability

reagent type: catalyst
core: vanadium

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

690 °C (lit.)

density

3.35 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

greener alternative category

SMILES string

O=[V](=O)O[V](=O)=O

InChI

1S/5O.2V

InChI key

GNTDGMZSJNCJKK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more details.

Application

Selective ammoxidation catalysts for such reactions as conversion of toluene to benzonitrile or propylene to acrylonitrile were prepared by a sol-gel method from V2O5, Sb2O3 and high purity hydrogen peroxide.[1] Preparation of new stabilized, oxide ion-conducting, bismuth vanadate phases by a microwave assisted method, fromV2O5, Bi2O3 and other solid oxides, was reported. These ceramics show promise in solid oxide fuel cells, water-vapor electrolyzers and oxygen sensors.[2]

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Carc. 1B - Lact. - Muta. 2 - Repr. 2 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Respiratory system, Respiratory Tract

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yeon Seok Kim et al.
Applied biochemistry and biotechnology, 168(5), 1143-1152 (2012-09-06)
The bactericidal activity of TiO(2) nanoparticles under visible light is very important in regards to its practical applications. In this paper, we synthesized vanadium-pentoxide-loaded TiO(2) nanoparticles (V(2)O(5)-TiO(2)) using a chemical vapor condensation method, followed by the impregnation method, and characterized
Junkal Gutierrez et al.
ChemSusChem, 5(12), 2323-2327 (2012-10-13)
A bacterial cellulose mat was used as a template for the fabrication of conductive photoswitchable hybrid nanopaper by the incorporation of sol-gel synthesized vanadium nanoparticles. The resulting nanopaper, prepared through a green pathway, was able to photoinduce a reversible color
Gongming Wang et al.
ACS nano, 6(11), 10296-10302 (2012-10-12)
Here we report a new strategy to improve the electrochemical stability of vanadium oxide electrodes for pseudocapacitors. Vanadium oxides are known to suffer from severe capacitance loss during charging/discharging cycling, due to chemical dissolution and ion intercalation/deintercalation-induced material pulverization. We
Hongchang Pang et al.
Chemical communications (Cambridge, England), 49(15), 1536-1538 (2013-01-17)
A template-free strategy is exploited to bottom-up synthesize yolk-shell vanadium oxide through a two-step spontaneous assembly of hydrolytically formed subunits in a one-pot process. The unique structured vanadium pentoxide exhibits excellent cathode performance for lithium ion batteries.
Xiaojiao Kang et al.
Nanoscale, 5(1), 253-261 (2012-11-17)
In this study, multifunctional poly(acrylic acid) modified lanthanide-doped GdVO(4) nanocomposites [PAA@GdVO(4): Ln(3+) (Ln = Yb/Er, Yb/Ho, Yb/Tm)] were constructed by filling PAA hydrogel into GdVO(4) hollow spheres via photoinduced polymerization. The up-conversion (UC) emission colors (green, red and blue) can

Questions

1–3 of 3 Questions  
  1. I would like to solicite if you could provide me with information on the particle size of the different presentations of the reagent(V2O5) hopefully between 5-10um for be used on solid state batteries. I look forward to your response.

    1 answer
    1. The particle size of this material is not determined on a lot to lot basis. However, historical data suggests a broad range of 10 - 250 mesh or 58 - 2000 um.

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service