Skip to Content
Merck
All Photos(2)

Key Documents

203742

Sigma-Aldrich

Manganese(II) nitrate hydrate

99.99% trace metals basis

Synonym(s):

Manganous dinitrate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Mn(NO3)2·xH2O
CAS Number:
Molecular Weight:
178.95 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
crystals and lumps
Pricing and availability is not currently available.

grade

for analytical purposes

Quality Level

Assay

99.99% trace metals basis

form

crystals and lumps

composition

Degree of hydration, 4-6

impurities

≤150.0 ppm Trace Metal Analysis

application(s)

battery manufacturing

storage temp.

2-8°C

SMILES string

O.[Mn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Mn.2NO3.H2O/c;2*2-1(3)4;/h;;;1H2/q+2;2*-1;

InChI key

HBTFASPVVFSRRI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Manganese(II) nitrate hydrate can be used:
  • To fabricate tetragonal LiMn2O4 as a dual-functional pseudocapacitor-battery electrode via an in-situ ion-exchange method.
  • As a safe, cost-effective, and efficient electrolyte for rapid energy storage applications like supercapacitors.
  • As a precursor to prepare hybrid catalyst to enhance contaminants poisoning tolerance in solid oxide fuel cell cathodes.
  • To prepare MnO2–CoO catalyst to fabricate opto-electronic humidity sensors.

Pictograms

Flame over circleExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Therese, G.H.A. Kamath, P.V.
Chemistry of Materials, 10, 3364-3364 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service