Skip to Content
Merck
All Photos(4)

Key Documents

203548

Sigma-Aldrich

Lanthanum(III) nitrate hexahydrate

99.999% trace metals basis

Synonym(s):

Nitric acid, lanthanum (III) salt, hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
La(NO3)3 · 6H2O
CAS Number:
Molecular Weight:
433.01
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
solid

Quality Level

Assay

99.999% trace metals basis

form

solid

reaction suitability

reagent type: catalyst
core: lanthanum

impurities

≤15.0 ppm Trace Rare Earth Analysis

mp

65-68 °C

SMILES string

[La+3].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/La.3NO3.6H2O/c;3*2-1(3)4;;;;;;/h;;;;6*1H2/q+3;3*-1;;;;;;

InChI key

GJKFIJKSBFYMQK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lanthanum(III) nitrate hexahydrate is a transition metal catalyst widely used in many organic transformations such as chemoselective deprotection of acetonides, acetylation of alcohols, phenols, and amines with acetic anhydride, and synthesis of α-amino nitriles. It can also be used as a precursor in the preparation of lanthanum-based nanoparticles.

Application

Lanthanum(III) nitrate hexahydrate can be used as a catalyst in the synthesis of:
  • α-aminonitriles by a one-pot three-component coupling of the carbonyl compound, amine, and TMSCN.
  • Acetals and geminal diacetates (acylals)from aldehydes.
  • Bis(indolyl) methanes under solvent‐free conditions.
It can also be used as a starting material for the electrochemical synthesis of LaMnO3 thin films and solvothermal synthesis of doped lanthanide oxysulfide (La2O2S).

Features and Benefits

  • Thermal stability
  • Low toxicity
  • Mild reaction condition
  • Chemoselectivity
  • Low cost

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Ox. Sol. 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Tyler Mefford et al.
Physical chemistry chemical physics : PCCP, 21(6), 3327-3338 (2019-01-29)
Perovskite oxides are active room-temperature bifunctional oxygen electrocatalysts in alkaline media, capable of performing the oxygen reduction reaction (ORR) and oxygen evolution reaction (OER) with lower combined overpotentials relative to their precious metal counterparts. However, their semiconducting nature necessitates the
Sang Eun Lee et al.
The Journal of investigative dermatology, 130(4), 1063-1072 (2009-11-27)
Low-frequency sonophoresis (LFS) has been shown to disrupt the structure of stratum corneum (SC) lipid bilayers and enhance SC permeability. In this study, we examined the penetration pathway of lanthanum nitrate (LaNO(3)) tracer in viable epidermis after combined treatment of
Liping Wei et al.
Microvascular research, 80(1), 158-165 (2010-01-19)
The PKC-beta inhibitor ruboxistaurin (RBX or LY333531) prevents diabetic renal and retinal microvascular complications. However, the effect of RBX on diabetic cardiac microvascular dysfunction is still unclear. In this study, we aimed to investigate the effects and mechanisms of RBX
Hartwig Wolburg et al.
Histochemistry and cell biology, 130(1), 127-140 (2008-03-15)
The olfactory ensheathing (glial) cells (OECs) have been identified to be useful candidate cells to support regeneration after being transplanted into injured fiber tracts of the central nervous system. We investigated by means of immunocytochemistry and freeze-fracturing the morphology and
Wen-Feng Zhao et al.
Dalton transactions (Cambridge, England : 2003), 41(33), 10091-10096 (2012-08-02)
The reaction between polyoxometalate (POM) [TBA](12)[WZn{Zn(H(2)O)}(2)(ZnW(9)O(34))(2)] (TBA = tetrabutyl ammonium) and lanthanide (Ln) nitrate (Ln = La, Eu and Tb) in a mixed solvent of CH(3)CN and DMF yielded three noncentrosymmetric diamondoid Ln-POM solid materials, {[Ln(2)(DMF)(8)(H(2)O)(6)][ZnW(12)O(40)]}·4DMF (Ln-POM; Ln = La

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service