Skip to Content
Merck
All Photos(1)

Documents

529303

Sigma-Aldrich

Iron(III) nitrate nonahydrate

≥99.999% trace metals basis

Synonym(s):

Ferric nitrate nonahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Fe(NO3)3 · 9H2O
CAS Number:
Molecular Weight:
404.00
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

≥99.999% trace metals basis

form

solid

impurities

≤10.0 ppm Trace Metal Analysis

mp

47 °C (lit.)

application(s)

battery manufacturing

SMILES string

O.O.O.O.O.O.O.O.O.[Fe+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Fe.3NO3.9H2O/c;3*2-1(3)4;;;;;;;;;/h;;;;9*1H2/q+3;3*-1;;;;;;;;;

InChI key

SZQUEWJRBJDHSM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

A variety of organically templated iron phosphates, of interest because of their rich crystal chemistry and their ability to absorb large molecules in their micropores,were prepared using this compound.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lii, K-H. et al.
Chemistry of Materials, 10, 2599-2599 (1998)
Mara Ghiazza et al.
Chemical research in toxicology, 24(1), 99-110 (2010-12-22)
The mechanism of enhancement/inhibition of quartz toxicity induced by iron is still unclear. Here the amount of iron on a fibrogenic quartz (Qz) was increased by wet impregnation (Fe(NO(3))(3) 0.67 and 6.7 wt %). X-ray diffraction (XRD), XRF diffuse reflectance
Metal-mediated cross-linking in the generation of a marine-mussel adhesive.
Mary J Sever et al.
Angewandte Chemie (International ed. in English), 43(4), 448-450 (2004-01-22)
Runjuan Wang et al.
Bioresource technology, 120, 290-294 (2012-07-10)
The study investigated the effects of two-step pretreatment on fermentable sugar production from corn stalk silage. In the first step, the corn stalk silage was extracted by tepid water and then the solid was pretreated using Fe(NO(3))(3) as catalyst. The
Marek Basler et al.
Proteomics, 6(23), 6194-6206 (2006-11-30)
Restricting bacterial growth by iron-chelating proteins that reduce iron availability in mucosal secretions and body fluids belongs to basic mechanisms of innate immunity. Most pathogens and commensals thus developed gene regulons responding to iron concentration and encoding iron acquisition systems

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service