Skip to Content
Merck
All Photos(1)

Key Documents

203696

Sigma-Aldrich

Magnesium nitrate hexahydrate

99.999% trace metals basis

Synonym(s):

Magnesium dinitrate hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Mg(NO3)2 · 6H2O
CAS Number:
Molecular Weight:
256.41
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

vapor density

6 (vs air)

Assay

99.999% trace metals basis

form

solid
solid

impurities

≤15.0 ppm Trace Metal Analysis

mp

89 °C (dec.) (lit.)

SMILES string

O.O.O.O.O.O.[Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Mg.2NO3.6H2O/c;2*2-1(3)4;;;;;;/h;;;6*1H2/q+2;2*-1;;;;;;

InChI key

MFUVDXOKPBAHMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Magnesium nitrate hexahydrate is an inorganic salt hydrate phase change material(PCM) widely used in thermal energy storage devices. It is the most preferred inorganic PCM because of its applicable phase change temperature of about 89 °C, incombustibility, ultra-high phase change enthalpy 162.8 Jg−1, and low cost. It can also be used as a precursor to synthesize MgO-based catalysts for various applications.

Application


  • Understanding the Roles of Excipients in Moisture Management in Solid Dosage Forms.: This study explores the use of excipients, including Magnesium nitrate hexahydrate, in managing moisture in solid dosage pharmaceutical forms. It highlights the role of Magnesium nitrate hexahydrate in enhancing the stability and efficacy of moisture-sensitive drugs (Veronica et al., 2024).

  • Printing Composites with Salt Hydrate Phase Change Materials for Thermal Energy Storage.: The research discusses the application of Magnesium nitrate hexahydrate in phase change materials for thermal energy storage, emphasizing its thermal properties and suitability for energy-efficient systems (Lak et al., 2023).

  • How do layered double hydroxides evolve-First in situ insights into their synthesis processes.: This paper provides insights into the synthesis of layered double hydroxides, with Magnesium nitrate hexahydrate playing a crucial role in the process. It examines the material′s properties and potential applications in various industries (Doungmo et al., 2022).

  • Encapsulation of hygroscopic liquids via polymer precipitation in non-aqueous emulsions.: The study investigates the encapsulation of hygroscopic liquids, using Magnesium nitrate hexahydrate as a key component to improve encapsulation efficiency and stability (Lak et al., 2022).

  • A Wide Bandgap Semiconducting Magnesium Hydrogel: Moisture Harvest, Iodine Sequestration, and Resistive Switching.: This research highlights the development of a magnesium hydrogel incorporating Magnesium nitrate hexahydrate, demonstrating its effectiveness in moisture harvesting and iodine sequestration, with potential applications in semiconductors (Alam et al., 2022).

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H R Shakur et al.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine, 118, 43-55 (2016-09-10)
In order to efficiently remove of uranium anionic species (which are the most dominant species of uranium in natural water at neutral pH) from contaminated waters, nano-NaX zeolite was synthesized and then modified using various divalent cations (Mg2+, Ca2+, Mn2+)
Mohamed Abdellatif Bensegueni et al.
Acta crystallographica. Section C, Structural chemistry, 71(Pt 3), 222-228 (2015-03-04)
Two alkaline earth-tetrazole compounds, namely catena-poly[[[triaquamagnesium(II)]-μ-5,5'-(azanediyl)ditetrazolato-κ(3)N(1),N(1'):N(5)] hemi{bis[μ-5,5'-(azanediyl)ditetrazolato-κ(3)N(1),N(1'):N(2)]bis[triaquamagnesium(II)]} monohydrate], {[Mg(C(2)HN(9))(H(2)O)(3)][Mg(2)(C(2)HN(9))(2)(H(2)O)(6)]0.5·H(2)O}n, (I), and bis[5-(pyrazin-2-yl)tetrazolate] hexaaquamagnesium(II), (C(5)H(3)N(6))[Mg(H(2)O)(6)], (II), have been prepared under hydrothermal conditions. Compound (I) is a mixed dimer-polymer based on magnesium ion centres and can be regarded as the first example
Zhaoying Wu et al.
Colloids and surfaces. B, Biointerfaces, 120, 38-46 (2014-06-07)
Mesoporous magnesium silicate (m-MS) was synthesized, and the in vitro degradability, bioactivity and primary cell responses to m-MS were investigated. The results suggested that the m-MS with mesoporous channels of approximately 5nm possessed the high specific surface area of 451.0m(2)/g
Raquel C B Marques et al.
Talanta, 129, 594-599 (2014-08-17)
Human epidermal growth factor receptor 2 (HER2) is a breast cancer biomarker that plays a major role in promoting breast cancer cell proliferation and malignant growth. The extracellular domain (ECD) of HER2 can be shed into the blood stream and
Mario Ledda et al.
Biomedical materials (Bristol, England), 10(3), 035005-035005 (2015-06-05)
In this work, titanium (Ti) supports have been coated with glass-ceramic films for possible applications as biomedical implant materials in regenerative medicine. For the film preparation, a pulsed laser deposition (PLD) technique has been applied. The RKKP glass-ceramic material, used

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service