Skip to Content
Merck
All Photos(3)

Key Documents

P5413

Sigma-Aldrich

Poly(ethylene glycol)

for molecular biology, average mol wt 8,000

Synonym(s):

Poly (ethylene oxide), Polyethylene glycol, PEG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H(OCH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.31

grade

for molecular biology

Quality Level

form

powder

mol wt

average mol wt 8,000

solubility

water: soluble (approximately 630 mg/ml, 20 °C)

density

1.0845 g/mL at 70 °C
1.0689 g/mL at 90 °C

foreign activity

DNase, RNase and NICKase, none detected

SMILES string

C(CO)O

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Polyethylene glycol (PEG) is a polymer of ethylene oxide and water, obtained upon condensation. PEGs are susceptible to oxidative degradation in the presence of air. Minimizing the exposure of PEG to elevated temperatures and/or exposure to oxygen or addition of an antioxidant can limit the amount of degradation. PEGs do not hydrolyze or deteriorate upon storage. PEGs do not support the growth of molds.
Polyethylene glycol (PEG) is used in detergents, emulsifying agents, soaps, metal costing process, pharmaceutical preparations and cosmetics. PEG is commonly used as a lubricant. It′s application is also observed in chromatographic stationary phases. PEG prevents crystal formation in emulsions.

Application

Poly(ethylene glycol) has been used:
  • in protoplast suspension
  • to prevent phase separation in emulsion
  • in phage tail-like particles precipitation
  • for purifying and intensifying products of polymerase chain reaction

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John M Murray
Journal of microscopy, 267(2), 193-213 (2017-03-23)
The ability to replace genes coding for cellular proteins with DNA that codes for fluorescent protein-tagged versions opens the way to counting the number of molecules of each protein component of macromolecular assemblies in vivo by measuring fluorescence microscopically. Converting
Associations between HLA class I alleles and escape mutations in the hepatitis B virus core gene in New Zealand-resident Tongans
Abbott William GH, et al.
Journal of Virology, 84(1), 621-629 (2010)
Genes for the biosynthesis of the fungal polyketides hypothemycin from Hypomyces subiculosus and radicicol from Pochonia chlamydosporia
Reeves CD
Applied and Environmental Microbiology, 74(16), 5121-5129 (2008)
Induction and Purification of C. difficile Phage Tail-Like Particles
Clostridium difficile, 167-175 (2016)
Retinol encapsulated into amorphous Ca2+ polyphosphate nanospheres acts synergistically in MC3T3-E1 cells
Muller Werner EG, et al.
European Journal of Pharmaceutics and Biopharmaceutics, 93, 214-223 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service