911879
NanoFabTx™ microfluidic - micro
device kit for synthesis of 10-30 μm particles
Synonym(s):
Microfluidic kit, Microparticle, NanoFabTx™ device kit, Nanoformulation
Sign Into View Organizational & Contract Pricing
All Photos(4)
About This Item
Recommended Products
description
Microfludic device kit for synthesizing 10-30 μm microparticles
Kit components :
Quality Level
application(s)
advanced drug delivery
Looking for similar products? Visit Product Comparison Guide
General description
NanoFabTx™ Microfluidic - micro device kit is designed to work with the NanoFabTx™ - micro reagent kits like (Cat. No. 912212) for the synthesis of 10-30 μm particles. The device kit includes a comprehensive protocol, pre-assembled microfluidic chip manifold, tubing, and required accessories and is compatible with either the Dolomite microfluidics system or a standard syringe pump (compatible microfluidics system or syringe pump required separately).
Application
NanoFabTx™ Microfluidic - micro device kit in combination with the NanoFabTx™ - micro reagent kits (Cat. No. 912212) can be used to create drug-encapsulating polymeric microparticles 10-30 μm in size. Because of the adjustable flow rate, small reaction window, and adjustable concentrations, microfluidic-based synthesis is a powerful tool for fabricating size-controlled particles to encapsulate drugs, small molecules, mRNA, DNA, and proteins. The chip geometries and flow parameters enable facile, scalable synthesis of monodispersed particles with consistent size and drug loading to facilitate rapid formulation screening.
Features and Benefits
- Simplifies polymer chemistry of microfluidic formulation development for enhancing drug bioavailability
- Accelerates optimization of drug encapsulation and loading efficiency in microparticles
- Provides step-by-step microfluidic-based protocols developed and tested by our formulation scientists
- Minimizes waste of expensive reagents
Legal Information
NANOFABTX is a trademark of Sigma-Aldrich Co. LLC
related product
Product No.
Description
Pricing
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Journal of nanobiotechnology, 16(1), 12-12 (2018-02-13)
The process of optimization and fabrication of nanoparticle synthesis for preclinical studies can be challenging and time consuming. Traditional small scale laboratory synthesis techniques suffer from batch to batch variability. Additionally, the parameters used in the original formulation must be
Genes, 9(2) (2018-02-22)
Microfluidic devices present unique advantages for the development of efficient drug carrier particles, cell-free protein synthesis systems, and rapid techniques for direct drug screening. Compared to bulk methods, by efficiently controlling the geometries of the fabricated chip and the flow
Advanced drug delivery reviews, 128, 101-114 (2017-12-27)
Microfluidic chips allow the rapid production of a library of nanoparticles (NPs) with distinct properties by changing the precursors and the flow rates, significantly decreasing the time for screening optimal formulation as carriers for drug delivery compared to conventional methods.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service