Skip to Content
Merck
All Photos(1)

Key Documents

193666

Sigma-Aldrich

Chloropentafluorobenzene

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6F5
CAS Number:
Molecular Weight:
202.51
Beilstein:
1819389
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.424 (lit.)

bp

122-123 °C/750 mmHg (lit.)

density

1.568 g/mL at 25 °C (lit.)

functional group

chloro
fluoro

SMILES string

Fc1c(F)c(F)c(Cl)c(F)c1F

InChI

1S/C6ClF5/c7-1-2(8)4(10)6(12)5(11)3(1)9

InChI key

KGCDGLXSBHJAHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Reaction between chloropentafluorobenzene and strong N-bases in polar aprotic solvents and in the presence of water has been investigated. Chloropentafluorobenzene on reaction with ammonia yields 4-chloro-2,3,5,6-tetrafluoroaniline and 2-chloro-3,4,5,6-tetrafluoroaniline.

Application

Chloropentafluorobenzene has been used in the preparation of:
  • 1,2-difluoro-1,2-bis-(pentafluorophenyl)dichlorane via fluorination with elemental fluorine at 128°C
  • 5-chloro-1-(difluorochloro)-2,3,4,5,6,6-hexafluoro-1,3-cyclohexadiene via reaction with chlorine trifluoride at −78°C

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

5-Chloro-1-(difluorochloro)-2, 3, 4, 5, 6, 6-hexafluoro-1, 3-cyclohexadiene: Synthesis and characterization.
Obaleye JA and Sams LC.
Journal of Fluorine Chemistry, 23(5), 479-479 (1983)
E R Kinkead et al.
Toxicology and industrial health, 7(4), 309-318 (1991-07-01)
Chloropentafluorobenzene (CPFB) has been identified as a candidate simulant for nonpersistent chemical warfare agents. Acute toxicity studies have shown that CPFB has limited adverse effects on laboratory animals. A 21-day inhalation study of rats and mice to 2.5, 0.8, and
Błazej Gierczyk et al.
The Journal of organic chemistry, 68(8), 3139-3144 (2003-04-12)
Products of reactions between chloropentafluorobenzene and strong N-bases (DBN, DBU, TBD, and MTBD) in polar aprotic solvents and in the presence of water were isolated and identified by analytical and spectroscopic methods. The products of these dehydrohalogenation reactions are apropriate
Synthesis of 1, 2 difluoro-1, 2-bis (pentafluorophenyl) dichlorane.
Obaleye JA and Sams LC.
Journal of Fluorine Chemistry, 24(4), 413-418 (1984)
J N McDougal et al.
Microscopy research and technique, 37(3), 214-220 (1997-05-01)
Skin samples from Fischer-344 rats and Hartley guinea pigs exposed dermally to 1,2-dichlorobenzene (DCB) and chloropentafluorobenzene (CPFB) for up to 4 h were examined for chemical-induced damage. Samples were stained with hematoxylin and eosin and scored for polymorphonuclear cell (PMN)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service