Skip to Content
Merck
All Photos(2)

Key Documents

442593

Sigma-Aldrich

Tris(pentafluorophenyl)borane

greener alternative

95%

Synonym(s):

Perfluorotriphenylboron

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6F5)3B
CAS Number:
Molecular Weight:
511.98
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

95%

form

powder

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

126-131 °C (lit.)

greener alternative category

SMILES string

Fc1c(F)c(F)c(B(c2c(F)c(F)c(F)c(F)c2F)c3c(F)c(F)c(F)c(F)c3F)c(F)c1F

InChI

1S/C18BF15/c20-4-1(5(21)11(27)16(32)10(4)26)19(2-6(22)12(28)17(33)13(29)7(2)23)3-8(24)14(30)18(34)15(31)9(3)25

InChI key

OBAJXDYVZBHCGT-UHFFFAOYSA-N

General description

B(C6F5)3 is a Lewis acid, stronger than the inorganic parent molecule BF3. In contrast to the prominent Lewis acids, the pentafluorides of the heavier elements of group 15, B(C6F5)3 has no oxidizing properties. B(C6F5)3 forms adducts with neutral and weaker Lewis bases. B(C6F5)3 has the capability to easily accept anionic ligands bonded to metals.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

A versatile reagent widely used in the preparation of d0 arene and other organometallic complexes useful as polymerization catalysts.
Used with tri-tert-butylphosphine (570958) to study the heterolytic cleavage of dihydrogen at room temperature and one atmosphere pressure.

Features and Benefits

A versatile reagent widely used in the preparation of d0 arene and other organometallic complexes useful as polymerization catalysts.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organometallics, 15, 3600-3600 (1996)
Facile heterolytic cleavage of dihydrogen by phosphines and boranes.
Gregory C Welch et al.
Journal of the American Chemical Society, 129(7), 1880-1881 (2007-01-31)
Berthold Wegner et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 7(17), 2001322-2001322 (2020-10-01)
Molecular doping allows enhancement and precise control of electrical properties of organic semiconductors, and is thus of central technological relevance for organic (opto-) electronics. Beyond single-component molecular electron acceptors and donors, organic salts have recently emerged as a promising class
Dan Voicu et al.
ChemSusChem, 8(24), 4202-4208 (2015-11-27)
Separation of gaseous olefins and paraffins is one of the most important separation processes in the industry. Development of new cost-effective technologies aims at reducing the high energy consumption during the separation process. Here, we took advantage of the reaction
Journal of the American Chemical Society, 118, 10008-10008 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service