O2706
Octafluorotoluene
98%
Synonym(s):
Perfluorotoluene
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C6F5CF3
CAS Number:
Molecular Weight:
236.06
Beilstein:
2057072
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
refractive index
n20/D 1.368 (lit.)
bp
104 °C (lit.)
density
1.666 g/mL at 25 °C (lit.)
SMILES string
Fc1c(F)c(F)c(c(F)c1F)C(F)(F)F
InChI
1S/C7F8/c8-2-1(7(13,14)15)3(9)5(11)6(12)4(2)10
InChI key
USPWUOFNOTUBAD-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
68.0 °F - closed cup
Flash Point(C)
20 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
M H Baker et al.
Biomedical & environmental mass spectrometry, 16(1-12), 211-213 (1988-10-01)
The use of octafluorotoluene (OFT) as a versatile gas chromatographic/mass spectrometric derivatizing agent for steroids containing alcoholic, phenolic or alpha, beta-unsaturated keto functions, is described. Two distinct derivatizing schemes can be utilized, involving (method 1) a phase-transfer reaction, employing a
Z Fang et al.
Anesthesia and analgesia, 83(5), 1097-1104 (1996-11-01)
We examined the anesthetic and convulsant properties of 16 unfluorinated to completely fluorinated aromatic compounds, having six to nine carbon atoms (e.g., benzene to 1,3,5-tris(trifluoromethyl)benzene), and four cycloalkanes (cyclopentane to cyclooctane). Benzene, fluorobenzene, toluene, p-xylene, ethylbenzene, and cyclopentane caused excitation
Alexander V Belyakov et al.
The journal of physical chemistry. A, 124(49), 10216-10224 (2020-11-18)
The equilibrium molecular structure of the gaseous fluorofullerene C60F36 has been determined for the first time by the electron diffraction method with the use of quantum chemical calculations up to the RI-MP2/def2-TZVPP level of theory. Vibrational amplitudes and quadratic and
Naohide Shinohara et al.
International journal of environmental research and public health, 7(9), 3348-3358 (2010-10-16)
A modified procedure was developed for the measurement of the effective air exchange rate, which represents the relationship between the pollutants emitted from indoor sources and the residents' level of exposure, by placing the dosers of tracer gas at locations
LUMOS--A Sensitive and Reliable Optode System for Measuring Dissolved Oxygen in the Nanomolar Range.
Philipp Lehner et al.
PloS one, 10(6), e0128125-e0128125 (2015-06-02)
Most commercially available optical oxygen sensors target the measuring range of 300 to 2 μmol L-1. However these are not suitable for investigating the nanomolar range which is relevant for many important environmental situations. We therefore developed a miniaturized phase
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service