Skip to Content
Merck
All Photos(1)

Documents

101052

Sigma-Aldrich

2,3,4,5,6-Pentafluorobenzyl bromide

99%

Synonym(s):

α-Bromo-2,3,4,5,6-pentafluorotoluene, (Bromomethyl)pentafluorobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6F5CH2Br
CAS Number:
Molecular Weight:
260.99
Beilstein:
647808
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

refractive index

n20/D 1.471 (lit.)

bp

174-175 °C (lit.)

mp

19-20 °C (lit.)

density

1.728 g/mL at 25 °C (lit.)

SMILES string

Fc1c(F)c(F)c(CBr)c(F)c1F

InChI

1S/C7H2BrF5/c8-1-2-3(9)5(11)7(13)6(12)4(2)10/h1H2

InChI key

XDEPVFFKOVDUNO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Derivatizing agent for GC analysis of polyfunctional thiols.
For the preparation of pentafluorobenzyl esters of organic acids for determination by capillary and GC. Also used to derivatize N-7-substituted guanine adducts of DNA for determination by GC-electron capture-MS.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

181.4 °F - closed cup

Flash Point(C)

83 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D H Fisher et al.
Journal of chromatography, 452, 51-60 (1988-10-28)
The following sequence of analytical steps was used to determine the amount of 5-methylcytosine (mol-%) in calf thymus and human lymphocyte DNA:acid hydrolysis of the DNA, derivatization (pentafluorobenzyl bromide, solid phase extraction, pivalic anhydride), internal standard addition, solid phase extraction
Laura Mateo-Vivaracho et al.
Journal of chromatography. A, 1121(1), 1-9 (2006-05-09)
A fast and automated method for the analysis at ng L(-1) level of aroma-powerful polyfunctional thiols has been developed and applied to wine. The sample is just poured in a 20 mL vial and its vapour extracted with a poly(dimethylsiloxane)-divinylbenzene
Journal of Chromatography A, 539, 232-232 (1991)
M Saha et al.
Journal of chromatography. A, 712(2), 345-354 (1995-10-13)
A 112-micrograms sample of DNA was spiked with 103 pg of N7-(2'-hydroxyethyl)guanine and 100 pg of N7-(2'-hydroxyethyl-d4)guanine, the internal standard. The sample was subjected to the following sequence of steps: heating at 100 degrees C, precipitation of the DNA with
Yan Li et al.
Frontiers in bioengineering and biotechnology, 7, 394-394 (2019-12-24)
The main challenges in multienzymatic cascade reactions for CO2 reduction are the low CO2 solubility in water, the adjustment of substrate channeling, and the regeneration of co-factor. In this study, metal-organic frameworks (MOFs) were prepared as adsorbents for the storage

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service