Skip to Content
Merck
All Photos(3)

Key Documents

191973

Sigma-Aldrich

Poly(2-acrylamido-2-methyl-1-propanesulfonic acid) solution

average Mw 2,000,000, 15 wt. % in H2O

Synonym(s):

2-Acrylamido-2-methyl-1-propanesulfonic acid polymer, 2-Acrylamido-2-methylpropanesulfonic acid polymer, PolyAMPS

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

mol wt

average Mw 2,000,000

concentration

15 wt. % in H2O

pH

1.0-3.0

surface tension

79.8 dyn/cm, 1 wt. %

viscosity

~200,000 cP, Brookfield RVT(lit.)

acid number

41‑45 mg KOH/g

SMILES string

CC(C)(CS(O)(=O)=O)NC(=O)C=C

InChI

1S/C7H13NO4S/c1-4-6(9)8-7(2,3)5-13(10,11)12/h4H,1,5H2,2-3H3,(H,8,9)(H,10,11,12)

InChI key

XHZPRMZZQOIPDS-UHFFFAOYSA-N

Application

Thickening agent for acidic or basic cleaning agents, friction reducing agent, water-based lubricant, mineral scale remover and suspension aid for pigments and fillers. Rheology control agent in water and some organic solvents.

Features and Benefits

Solutions are pseudoplastic but not thixotropic. Stable in 50% HCl solutions. Compatible with anionic and non-ionic thickeners, surfactants and preservatives.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ryusei Imabuchi et al.
BMC musculoskeletal disorders, 12, 213-213 (2011-10-01)
We have recently found a phenomenon that spontaneous regeneration of a hyaline cartilage-like tissue can be induced in a large osteochondral defect by implanting a double-network (DN) hydrogel plug, which was composed of poly-(2-Acrylamido-2-methylpropanesulfonic acid) and poly-(N, N'-Dimetyl acrylamide), at
Munehiro Ogawa et al.
Journal of biomedical materials research. Part A, 100(9), 2244-2251 (2012-04-12)
The purpose of this study was to determine the in vivo cartilage induction effect of the poly(2-acrylamido-2-methylpropanesulfonic acid) (PAMPS) single-network (SN) gel and poly(N,N'-dimethyl acrylamide) (PDMAAm) SN gel in comparison with that of the PAMPS/PDMAAm double-network (DN) gel. An osteochondral
Kazunobu Arakaki et al.
Journal of biomedical materials research. Part A, 93(3), 1160-1168 (2009-09-22)
This study evaluated the in vivo influence of a poly-(2-Acrylamido-2-methylpropane sulfonic acid)/poly-(N,N'-dimetyl acrylamide) (PAMPS/PDMAAm) double-network (DN) hydrogel on counterface cartilage in rabbit knee joints and its ex vivo friction properties on normal cartilage. In the first experiment, the DN gel
Yoshie Tanabe et al.
Journal of materials science. Materials in medicine, 19(3), 1379-1387 (2007-10-05)
The study evaluated biological reaction of four types of novel double network gels in muscle and subcutaneous tissues, using implantation tests according to the international guideline. The implantation tests demonstrated that, although poly (2-acrylamide-2-metyl-propane sulfonic acid)/poly (N,N'-dimetyl acrylamide) (PAMPS/PDMAAm) gel
Nobuto Kitamura et al.
The American journal of sports medicine, 39(6), 1160-1169 (2011-04-05)
A double-network (DN) gel, which was composed of poly-(2-acrylamido-2-methylpropanesulfonic acid) and poly-(N,N'-dimetyl acrylamide) (PAMPS/PDMAAm), has the potential to induce chondrogenesis both in vitro and in vivo. To establish the efficacy of a therapeutic strategy for an articular cartilage defect using

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service