190888
Poly[(2-ethyldimethylammonioethyl methacrylate ethyl sulfate)-co-(1-vinylpyrrolidone)]
average Mw <1,000,000 by GPC, 20 wt. % in H2O
Synonym(s):
Poly(1-vinylpyrrolidone-co-2-dimethylaminoethyl methacrylate), Polyquaternium D 11
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
form
solid
mol wt
average Mw <1,000,000 by GPC
concentration
20 wt. % in H2O
density
1.05 g/mL at 25 °C
SMILES string
[S](=O)(=O)(OCC)OCC.N1(CCCC1=O)C=C.N(CCOC(=O)C(=C)C)(C)C
InChI
1S/C8H15NO2.C6H9NO.C4H10O4S/c1-7(2)8(10)11-6-5-9(3)4;1-2-7-5-3-4-6(7)8;1-3-7-9(5,6)8-4-2/h1,5-6H2,2-4H3;2H,1,3-5H2;3-4H2,1-2H3
InChI key
XXJPSODUGQZOHA-UHFFFAOYSA-N
Related Categories
General description
Poly[(2-ethyldimethylammonioethyl methacrylate ethyl sulfate)-co-(1-vinylpyrrolidone)] is a cationic polyelectrolyte that can be used as a reducing and protective agent to synthesize nanoparticles.
Preparation Note
Quaternized with diethyl sulfate
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service