Skip to Content
Merck
All Photos(1)

Documents

181463

Sigma-Aldrich

Polyisobutylene

average Mw ~1,000,000, average Mn ~600,000 by GPC/MALLS, average Mv ~1,200,000

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2C(CH3)2]n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

solid (or Chunk(s))

mol wt

average Mn ~600,000 by GPC/MALLS
average Mv ~1,200,000
average Mw ~1,000,000

contains

500 ppm BHT as stabilizer

color

colorless, or white

refractive index

n20/D 1.51

density

0.92 g/mL at 25 °C (lit.)

SMILES string

CC(C)=C

InChI

1S/C4H8/c1-4(2)3/h1H2,2-3H3

InChI key

VQTUBCCKSQIDNK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • The use of polyisobutylene-based polymers in ophthalmology: Discusses the application of polyisobutylene-based polymers in ophthalmologic devices, highlighting its potential in biomedical applications (L Pinchuk, 2022).
  • Synthesis of a polyisobutylene-tagged fac-Ir (ppy) 3 complex and its application as recyclable visible-light photocatalyst in a continuous flow process: Presents a novel use of polyisobutylene as a support for photocatalysts, enhancing recyclability and efficiency in light-driven chemical reactions (D Rackl, P Kreitmeier, O Reiser, 2016).
  • Polyisobutylene-based thermoplastic elastomers for manufacturing polymeric heart valve leaflets: In vitro and in vivo results: Explores the use of polyisobutylene-based elastomers in the production of heart valve leaflets, assessing their physical properties and compatibility (E Ovcharenko et al., 2019).
  • Polyisobutylene—new opportunities for medical applications: Reviews the potential of polyisobutylene in various medical applications, focusing on its properties and benefits for healthcare materials (D Barczikai et al., 2021).
  • Homogeneous and heterogeneous catalysts for the synthesis of highly reactive polyisobutylene: discovery, development and perspectives: Discusses advancements in catalyst technology for producing high-reactivity polyisobutylene, crucial for high-performance materials (IV Vasilenko, SV Kostjuk, 2021).

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yanfei Yang et al.
Journal of the American Chemical Society, 134(36), 14714-14717 (2012-08-29)
The studies described here show that a relatively low molecular weight, narrow polydispersity polyethylene (PE) wax (Polywax) can serve as a nontoxic and nonvolatile alternative to alkane solvents in monophasic catalytic organic reactions where catalysts and products are separated under
Corinne Vebert-Nardin
Chimia, 65(10), 782-786 (2011-11-08)
With this article, we wish to give an overview of our main research activities assessing the potential of a suitable polymer modification of DNA fragments to self-assemble biologically active nanostructures. Specifically, the grafting of a hydrophobic polymer segment on DNA
Judit E Puskas et al.
Biomaterials, 31(9), 2477-2488 (2009-12-26)
This paper presents the synthesis and characterization of a polyisobutylene (PIB)-based nanostructured carbon-reinforced thermoplastic elastomer. This thermoplastic elastomer is based on a self-assembling block copolymer having a branched PIB core carrying -OH functional groups at each branch point, flanked by
Deniz C Tuncaboylu et al.
Langmuir : the ACS journal of surfaces and colloids, 26(10), 7574-7581 (2010-01-29)
Organic-inorganic hybrid materials attract particular interest because of their excellent mechanical properties. Here, we report the synthesis of hybrid cryogels consisting of interpenetrated polyisobutylene and silica networks. The gels were prepared by cross-linking of butyl rubber in cyclohexane containing silica
Hong Seok Choi et al.
Langmuir : the ACS journal of surfaces and colloids, 28(1), 849-854 (2011-11-25)
The optical properties of polymeric materials, such as transmission loss and the thermo-optic coefficient, determine their utility in numerous applications, ranging from nanotechnology to the automotive and aerospace industries. However, because of the wide variation in the physical properties of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service