Skip to Content
Merck
All Photos(1)

Key Documents

388696

Sigma-Aldrich

Polybutenes

average Mn ~920 by VPO, isobutylene >90 %

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH2C(CH3)2-]x[-CH2CH(C2H5)-]y
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

liquid

mol wt

average Mn ~920 by VPO

composition

isobutylene, >90%

viscosity

200-235 cSt(100 °C)(lit.)

transition temp

pour point (ASTM D 97) −7 °C

density

0.89 g/mL at 25 °C

SMILES string

CCC=C.C\C=C\C

InChI

1S/2C4H8/c2*1-3-4-2/h3-4H,1-2H3;3H,1,4H2,2H3/b4-3+;

InChI key

WTOOLIQYCQJDBG-BJILWQEISA-N

Looking for similar products? Visit Product Comparison Guide

Application

Hydrophobic material is derivatized to make detergents and anti-rust additives. Adds tack, flexibility and water repellency to adhesives and coatings. Contributes ′cling′ to LLDPE films. Plasticizer for polymers, lubricants and metal-working fluids.

Features and Benefits

Tackiness increases with molecular weight. Some internal double bonds may be present.

Physical form

Terminal vinyl group.

Other Notes

remainder 1- and 2-butenes

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel Brassard et al.
Lab on a chip, 11(23), 4099-4107 (2011-11-02)
Microfluidics has emerged as a valuable tool for the high-resolution patterning of biological probes on solid supports. Yet, its widespread adoption as a universal biological immobilization tool is still limited by several technical challenges, particularly for the patterning of isolated
Anders Höglund et al.
Macromolecular bioscience, 12(2), 260-268 (2011-11-19)
The pre-polymer poly(but-2-ene-1,4-diyl malonate) (PBM) and a series of PBM-based materials are shown to be degradable under physiological conditions in vitro and they are therefore presented as potential materials for biomedical applications. Four different PBM-based materials are synthesized: a PBM
M Risbud et al.
Biomaterials, 22(12), 1591-1597 (2001-05-26)
The aim of the study was to prepare composites of poly(butylene terepthalate)/wollastonite (PBT/W), evaluate their properties and in vitro biocompatibility. Composites of PBT with wollastonite in two different proportions, viz. 70/30 (PW-30), 50/50 (PW-50) were prepared. The DSC studies indicate
Ibrahim Bulent Buttanri et al.
Ophthalmic plastic and reconstructive surgery, 29(2), 98-100 (2012-12-14)
To evaluate the effect of suturing the silicone rod to the tarsal plate and the suture material used in suturing on surgical success of frontalis suspension for congenital ptosis. Eighty eyes of 56 consecutive patients who underwent frontalis suspension surgery
Takanori Tanino et al.
Applied microbiology and biotechnology, 82(1), 59-66 (2008-11-06)
A Candida antarctica lipase B (CALB)-displaying yeast whole-cell biocatalyst was constructed with the integration of the CALB cell-surface display expression cassette in the yeast genome and cell fusion by mating. Lipase hydrolytic activity of the yeast whole-cell biocatalyst subsequently increased

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service