Skip to Content
Merck
All Photos(3)

Key Documents

P3683

Sigma-Aldrich

Paraplast Plus®

for tissue embedding

Synonym(s):

Paraffin – polyisobutylene mixture

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12171500
NACRES:
NA.47

Quality Level

form

solid

mp

56-57 °C (lit.)
56-57 °C

application(s)

hematology
histology

storage temp.

room temp

General description

N/A

Application

Paraplast Plus® has been used
  • in the examination of vertebral specimens of Malacoraja senta.
  • in the identification and expression analysis of PIN genes in rice.
  • in the study of glucose transport mechanisms in uterine tissues during pregnancy.
  • to embed Bouin′s fixed carp tissues, rat ventral prostatic fragments, and common bean (Phaseolus vulgaris L.) nodules.

Legal Information

Paraplast Plus is a registered trademark of Leica Biosystems

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification and expression analysis of PIN genes in rice
Miyashita, et al.
Plant Science, 178(5) (2010)
Nozomu Yoshioka et al.
Disease models & mechanisms, 13(5) (2020-06-03)
Loss-of-function mutations in dystonin (DST) can cause hereditary sensory and autonomic neuropathy type 6 (HSAN-VI) or epidermolysis bullosa simplex (EBS). Recently, DST-related diseases were recognized to be more complex than previously thought because a patient exhibited both neurological and skin
Micro-morphology of common bean (Phaseolus vulgaris L.)
nodules undergoing senescence
Fabia'n Ferna'ndez-Luquen?o ? L.
Acta Physiologiae Plantarum / Polish Academy of Sciences, Committee of Plant Physiology Genetics and Breeding (2008)
Shuaibin Zhang et al.
Scientific reports, 6, 23173-23173 (2016-03-19)
Early fruit development is crucial for crop production in tomato. After fertilization, the ovary undergoes cell division and cell expansion before maturation. Although the roles of regulatory signals such as hormone and carbohydrate during early fruit development have been studied
Harmful effects of carbamazepine on the postnatal development of the rat ventral prostate
Samara U Oliva
Reproductive Biology and Endocrinology (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service