Skip to Content
Merck
All Photos(2)

Key Documents

I13901

Sigma-Aldrich

Isoborneol

95%

Synonym(s):

(±)-Isoborneol, 2-exo-Bornyl alcohol, DL-Isoborneol, Isobornyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
EC Number:
MDL number:
UNSPSC Code:
12352212
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

95%

mp

212-214 °C (subl.) (lit.)

SMILES string

[H][C@@]12CC[C@@](C)([C@H](O)C1)C2(C)C

InChI

1S/C10H18O/c1-9(2)7-4-5-10(9,3)8(11)6-7/h7-8,11H,4-6H2,1-3H3/t7-,8-,10+/m1/s1

InChI key

DTGKSKDOIYIVQL-MRTMQBJTSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1 - Skin Irrit. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 2

Flash Point(F)

200.1 °F - closed cup

Flash Point(C)

93.4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

S P Bhatia et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 46 Suppl 11, S182-S184 (2008-07-22)
A toxicologic and dermatologic review of isoborneol when used as a fragrance ingredient is presented.
Rong Zhang et al.
European journal of drug metabolism and pharmacokinetics, 38(3), 159-169 (2013-04-17)
Borneol is a commonly used herbal medication in China and Japan. Previous studies have indicated that borneol could reduce the plasma concentrations of oneself and concomitant drugs, and its first-pass metabolism could be catalyzed by the cytochrome P450 3A (CYP3A)
Jianyu Su et al.
Journal of food science, 77(6), C658-C664 (2012-05-16)
The aims of this study were to optimize the preparation conditions of natural borneol/β-cyclodextrin (NB/β-CD) inclusion complex by ultrasound method, and to investigate its improvement of stability and solubility. The complex was characterized by different various spectroscopic techniques including Fourier
Jing-yi Zhao et al.
Journal of Zhejiang University. Science. B, 13(12), 990-996 (2012-12-12)
Borneol, a monoterpenoid alcohol, is used widely, particularly in combined formulas for preventing and curing cardiovascular and cerebrovascular diseases in traditional Chinese medicine. In order to understand the blood and brain pharmacokinetics after intravenous, intranasal, or oral administration and to
Eva Horváthová et al.
Mutagenesis, 27(5), 581-588 (2012-05-01)
Experimental evidences suggest that most essential oils possess a wide range of biological and pharmacological activities that may protect tissues against oxidative damage. In this study, we investigated DNA-protective effect of borneol, a component of many essential oils, against oxidative

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service