Skip to Content
Merck
All Photos(1)

Key Documents

02070595

Camphor (dl)

primary reference standard

Synonym(s):

(±)-Camphor, 1,7,7-Trimethylbicyclo[2.2.1]heptan-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
Beilstein:
1907611
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

primary reference standard

vapor density

5.2 (vs air)

vapor pressure

4 mmHg ( 70 °C)

shelf life

limited shelf life, expiry date on the label

expl. lim.

3.5 %

manufacturer/tradename

HWI

bp

204 °C (lit.)

mp

175-177 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

SMILES string

[H][C@](CC1=O)(CC2)C(C)(C)[C@]12C

InChI

1S/C10H16O/c1-9(2)7-4-5-10(9,3)8(11)6-7/h7H,4-6H2,1-3H3/t7-,10+/m1/s1

InChI key

DSSYKIVIOFKYAU-XCBNKYQSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Application

Reference Standard in the analysis of herbal medicinal products

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Sol. 2 - Skin Irrit. 2 - STOT SE 2 Inhalation

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 1

Flash Point(F)

147.9 °F - closed cup

Flash Point(C)

64.4 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guofeng Shang et al.
Journal of the Air & Waste Management Association (1995), 62(8), 873-879 (2012-08-25)
The characteristics and mechanisms of hydrogen sulfide (H2S) adsorption on a biochar through pyrolysis at various temperatures (100 to 500 degrees C) were investigated. The biochar used in the current study was derived from the camphor tree (Cinnamomum camphora). The
Yali V Zhang et al.
Nature neuroscience, 16(10), 1468-1476 (2013-09-10)
Animals tend to reject bitter foods. However, long-term exposure to some unpalatable tastants increases acceptance of these foods. Here we show that dietary exposure to an unappealing but safe additive, camphor, caused the fruit fly Drosophila melanogaster to decrease camphor
I Jiménez-Díaz et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 936, 80-87 (2013-09-06)
UV-filters are widely used in many personal care products and cosmetics. Recent studies indicate that some organic UV-filters can accumulate in biota and act as endocrine disruptors, but there are few studies on the occurrence and fate of these compounds
Dong-Bin Dang et al.
Chemical communications (Cambridge, England), 49(22), 2243-2245 (2013-02-12)
A series of 3D homochiral manganese-lanthanide frameworks have been synthesized based on chiral camphoric acid. In the heterometallic features, D-camphoric acids are unprecedentedly embedded in three coordination modes.
Skrollan Stockinger et al.
Journal of chromatography. A, 1269, 346-351 (2012-08-23)
Novel 3-(perfluoroalkanoyl)-(1R)-camphorate nickel complexes immobilized to poly(dimethylsiloxane) phases are presented. Immobilized 3-(perfluoroalkanoyl)-(1R)-camphorate nickel complexes with a trifluoromethyl (CF(3); nickel(II)-bis[(1R,4S)-3-trifluoromethanoyl-10-propylenoxycamphor]-polysiloxane Ni(tfpc)(2)@PS) and a heptafluoropropyl-substituent (C(3)F(7); nickel(II)-bis[(1R,4S)-3-heptafluorobutanoyl-10-propylenoxycamphor]-polysiloxane Ni(hfpc)(2)@PS) were synthesized, characterized and immobilized to polysiloxane. Ni(hfpc)(2)@PS was immobilized with a selector content

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service