Skip to Content
Merck
All Photos(2)

Documents

144657

Sigma-Aldrich

Trifluoroacetamide

97%

Synonym(s):

2,2,2-Trifluoroacetamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3CONH2
CAS Number:
Molecular Weight:
113.04
Beilstein:
1753625
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

bp

162.5 °C (lit.)

mp

65-70 °C (lit.)

SMILES string

NC(=O)C(F)(F)F

InChI

1S/C2H2F3NO/c3-2(4,5)1(6)7/h(H2,6,7)

InChI key

NRKYWOKHZRQRJR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Trifluoroacetamide is a good quencher of tryptophan fluorescence.

Application

Trifluoroacetamide was used as probe for determination of membrane potential and extra/intracellular volume of erythrocytes by fluorine-19 NMR studies.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Keita Tamura et al.
Plant & cell physiology, 58(5), 874-884 (2017-04-04)
The roots of Platycodon grandiflorus are widely used as a crude drug. The active components include a variety of triterpenoid saponins. Recent studies have revealed that Cyt P450 monooxygenases (P450s) function as triterpene oxidases in triterpenoid saponin biosynthesis in many
Chengquan Tan et al.
Scientific reports, 8(1), 10712-10712 (2018-07-18)
Sow milk contains necessary nutrients for piglets; however, the relationship between the levels of metabolites in sow milk and lactation performance has not been thoroughly elucidated to date. In this study, we analysed the metabolites in sow milk from Yorkshire
Xiao Yang et al.
Frontiers in plant science, 8, 2098-2098 (2018-01-13)
Glycine, the simplest amino acid in nature and one of the most abundant free amino acids in soil, is regarded as a model nutrient in organic nitrogen studies. To date, many studies have focused on the uptake, metabolism and distribution
Melody Anne de Laat et al.
PloS one, 13(6), e0200070-e0200070 (2018-06-30)
Hyperinsulinemia is a major risk factor for equine laminitis, a debilitating and painful foot condition. Sweet taste receptor (T1R2/3) inhibitors have been used to reduce the insulin and glucose responses to oral carbohydrates in other species. However, their effect in
Marc Galland et al.
Frontiers in plant science, 8, 1984-1984 (2017-12-08)
Although rice is a key crop species, few studies have addressed both rice seed physiological and nutritional quality, especially at the tissue level. In this study, an exhaustive "multi-omics" dataset on the mature rice seed was obtained by combining transcriptomics

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service