178721
(Diacetoxyiodo)benzene
98%
Synonym(s):
Iodobenzene I,I-diacetate, Iodosobenzene I,I-diacetate
About This Item
Recommended Products
Quality Level
Assay
98%
form
powder
reaction suitability
reagent type: catalyst
reagent type: oxidant
reaction type: C-H Activation
mp
161-163 °C (lit.)
functional group
iodo
SMILES string
CC(OI(OC(C)=O)C1=CC=CC=C1)=O
InChI
1S/C10H11IO4/c1-8(12)14-11(15-9(2)13)10-6-4-3-5-7-10/h3-7H,1-2H3
InChI key
ZBIKORITPGTTGI-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service