Skip to Content
Merck
All Photos(3)

Key Documents

482285

Sigma-Aldrich

Rhodium(II) acetate dimer

99.9% trace metals basis

Synonym(s):

Dirhodium tetraacetate, Tetrakis(acetato)dirhodium(II)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Rh2(OOCCH3)4
CAS Number:
Molecular Weight:
441.99
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.9% trace metals basis

form

powder

reaction suitability

reaction type: C-H Activation
reagent type: catalyst

SMILES string

CC(=O)O[Rh]OC(C)=O.CC(=O)O[Rh]OC(C)=O

InChI

1S/4C2H4O2.2Rh/c4*1-2(3)4;;/h4*1H3,(H,3,4);;/q;;;;2*+2/p-4

InChI key

SYBXSZMNKDOUCA-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for Rh-mediated C-H activation
Effective catalyst for ylide formation.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rhodium carbenoid mediated C-H activation of a tertiary methyl group: an enantiospecific approach to the angular triquinanes norsilphiperfolane and norcameroonanes
Srikrishna, A.; et al.
Synlett, 16, 2343-2346 (2011)
Jaroslav V Burda et al.
Journal of inorganic biochemistry, 102(1), 53-62 (2007-08-19)
In our study, we have determined the thermodynamic behavior for the replacement reaction of one and two acetyl-ligands from the diaqua-tetrakis(mu-acetylato)dirhodium(II,II) complex by purine DNA bases. The complexes were optimized at the density functional theory (DFT) level with the B3LYP
DNA cleavage by photogenerated Rh(2)(O(2)CCH(3))(4)(H(2)O)(2)(+).
P K Fu et al.
Inorganic chemistry, 40(11), 2476-2477 (2001-05-15)
Vânia André et al.
Acta crystallographica. Section C, Crystal structure communications, 64(Pt 10), m345-m348 (2008-10-08)
In this paper, we compare and discuss the very different crystal structures and supramolecular arrangements obtained when using different crystallization solvents with the same organometallic moiety. The new title tetrahydrofuran (THF) solvate, [Rh(2)(C(2)H(3)O(2))(4)(C(27)H(36)N(2))(2)] x 4 C(4)H(8)O, is compared with the
A Barnea et al.
Journal of inorganic biochemistry, 40(2), 103-110 (1990-10-01)
We have previously demonstrated that hypothalmic slices obtained from adult male rats accumulate 67Cu by two ligand-dependent, saturable processes: a high and low affinity process. To further establish the generality of these uptake processes, we defined the ligand requirements and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service