Skip to Content
Merck
All Photos(1)

Documents

13980

Sigma-Aldrich

Benzyltrimethylammonium chloride solution

technical, ~60% in H2O

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2N(Cl)(CH3)3
CAS Number:
Molecular Weight:
185.69
Beilstein:
3917255
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
NACRES:
NA.22

grade

technical

concentration

~60% in H2O

refractive index

n20/D 1.470

density

1.072 g/mL at 20 °C

SMILES string

[Cl-].C[N+](C)(C)Cc1ccccc1

InChI

1S/C10H16N.ClH/c1-11(2,3)9-10-7-5-4-6-8-10;/h4-8H,9H2,1-3H3;1H/q+1;/p-1

InChI key

KXHPPCXNWTUNSB-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Muta. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Sangeetha Vedula et al.
Archives of biochemistry and biophysics, 466(2), 260-266 (2007-08-07)
Trichodiene synthase is a terpenoid cyclase that catalyzes the cyclization of farnesyl diphosphate (FPP) to form the bicyclic sesquiterpene hydrocarbon trichodiene (89%), at least five sesquiterpene side products (11%), and inorganic pyrophosphate (PP(i)). Incubation of trichodiene synthase with 2-fluorofarnesyl diphosphate
Lena Edström et al.
Journal of chromatography. A, 1218(15), 1966-1973 (2010-10-05)
It has recently been demonstrated, using mathematical models, how peculiar overloaded band profiles of basic compounds are due to the local pH in the column when using low capacity buffers. In this study, overloaded peak shapes resulting after injection of
Weisheng Lin et al.
Analytica chimica acta, 583(1), 98-102 (2007-03-28)
The importance of benzyltriethyl ammonium chloride (BTEAC) in industrial applications has stimulated the development of a number of methods for its determination. In this paper, a high performance capillary electrophoresis (CE) method, coupled with an extraction technique for determining BTEAC
Purushothaman Gopinath et al.
The Journal of organic chemistry, 74(16), 6291-6294 (2009-07-22)
An efficient protocol is reported for the synthesis of thioesters from carboxylic acids with use of acyloxy phosphonium salts as intermediates and benzyltriethylammonium tetrathiomolybdate as the sulfur transfer reagent.
Nicola H Davies et al.
Journal of chromatography. A, 1119(1-2), 11-19 (2006-01-03)
The retention and overloading properties for eight basic solutes and two quaternary ammonium compounds were studied over the pH range 2.7-10.0 using phosphate and carbonate buffers. At low pH, a hybrid inorganic-organic silica-ODS phase (XTerra RP-18, 15 cm x 0.46

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service