Skip to Content
Merck
All Photos(4)

Key Documents

B8879

Sigma-Aldrich

Benzethonium chloride

≥97% (titration), ≥98% (HPLC)

Synonym(s):

(Diisobutylphenoxyethoxyethyl)dimethylbenzylammonium chloride, Phemerol chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H42ClNO2
CAS Number:
Molecular Weight:
448.08
Beilstein:
3898548
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

description

cationic

Assay

≥97% (titration)
≥98% (HPLC)

form

powder

mol wt

448.08 g/mol

mp

162-164 °C (lit.)

solubility

H2O: 100 mg/mL, clear, colorless

application(s)

microbiology

SMILES string

[Cl-].CC(C)(C)CC(C)(C)c1ccc(OCCOCC[N+](C)(C)Cc2ccccc2)cc1

InChI

1S/C27H42NO2.ClH/c1-26(2,3)22-27(4,5)24-13-15-25(16-14-24)30-20-19-29-18-17-28(6,7)21-23-11-9-8-10-12-23;/h8-16H,17-22H2,1-7H3;1H/q+1;/p-1

InChI key

UREZNYTWGJKWBI-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

The benzethonium chloride is a well characterized skin irritant and rare sensitizer.

Application

Benzethonium chloride has been used in a study to assess the minimum inhibitory concentrations of meticillin-resistant Staphylococcus aureus (MRSA) isolates from Malaysia. It has also been used in a study to prepare biocomposite films from sodium alginate and modified clay.

Disclaimer

The product is not intended for use as a biocide under global biocide regulations, including but not limited to US EPA′s Federal Insecticide Fungicide and Rodenticide Act, European Biocidal Products Regulation, Canada’s Pest Management Regulatory Agency, Turkey’s Biocidal Products Regulation, Korea’s Consumer Chemical Products and Biocide Safety Management Act (K-BPR) and others.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael J Orsini et al.
Journal of medicinal chemistry, 50(3), 462-471 (2007-02-03)
Metastin, also known as KiSS-1, the cognate ligand for the metastin receptor GPR54, is a peptide known to dramatically reduce metastasis in experimental models. Despite this, there is no reported structure for metastin nor any small molecule modulators of metastin
Elisabeth Lang et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 28(2), 347-354 (2011-08-26)
Benzethonium, an antimicrobial surfactant widely used as preservative of pharmaceuticals, topical wound care product and oral disinfectant, triggers apoptosis of several cell types. The apoptosis is preceded and possibly triggered by mitochondrial depolarization. Even though lacking mitochondria, erythrocytes may similarly
T Ichikawa et al.
Journal of dentistry, 36(11), 965-968 (2008-09-10)
The purpose of this study was to evaluate if the sugar alcohols erythritol, xylitol and sorbitol enhance the fungicidal effect of benzethonium chloride (BTC) toward in vitro candidal biofilms. An in vitroCandida albicans biofilm was formed on a plastic coverslip
Naoto Kojima et al.
Bioorganic & medicinal chemistry, 18(24), 8630-8641 (2010-11-16)
The convergent synthesis of fluorescence-labeled solamin, an antitumor Annonaceous acetogenin, was accomplished by two asymmetric alkynylations of 2,5-diformyl tetrahydrofuran with an alkyne tagged with fluorescent groups and another alkyne with an α,β-unsaturated γ-lactone. Assay for the growth inhibitory activity against
M N Shamsudin et al.
The Journal of hospital infection, 81(3), 206-208 (2012-05-29)
The minimum inhibitory concentrations (MICs) of 60 meticillin-resistant Staphylococcus aureus (MRSA) isolates from Malaysia to three antiseptic agents - benzalkonium chloride (BZT), benzethonium chloride (BAC) and chlorhexidine digluconate (CHG) - were determined. All isolates had MICs ranging from 0.5 to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service