138339
N-(2-Hydroxyethyl)phthalimide
99%
Synonym(s):
2-Phthalimidoethanol
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C10H9NO3
CAS Number:
Molecular Weight:
191.18
Beilstein:
147214
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
99%
form
solid
mp
126-128 °C (lit.)
functional group
hydroxyl
imide
SMILES string
OCCN1C(=O)c2ccccc2C1=O
InChI
1S/C10H9NO3/c12-6-5-11-9(13)7-3-1-2-4-8(7)10(11)14/h1-4,12H,5-6H2
InChI key
MWFLUYFYHANMCM-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
N-(2-Hydroxyethyl)phthalimide is the precursor for chloromethyl ethers used in the synthesis of purine acyclic nucleosides.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
J L Kelley et al.
Journal of medicinal chemistry, 24(12), 1528-1531 (1981-12-01)
A number of nitrogen analogues of 9-[(2-hydroxyethoxy)methyl]guanine [acylovir, Zovirax] containing amine functions in the side chain were synthesized and tested for antiviral activity. These purine acyclic nucleosides were prepared by reaction of tris(trimethylsilyl)guanine or 2,6-diaminopurine sodium salt with the chloromethyl
Molluscicidal activity of synthetic derivatives of 2-hydroxyethylphthalimide.
J N Lopes et al.
Revista brasileira de biologia, 40(3), 635-637 (1980-08-01)
Xuanrong Guo et al.
Biomacromolecules, 20(9), 3464-3474 (2019-07-25)
We report the design of reactive and hydrolytically degradable multilayers by the covalent layer-by-layer assembly of an azlactone-containing polymer, poly(2-vinyl-4,4-dimethylazlactone), with an acid-degradable, acetal-containing, small-molecule diamine linker. This approach yields cross-linked multilayers that contain (i) residual azlactone reactivity that can
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service