Skip to Content
Merck
All Photos(2)

Key Documents

185558

Sigma-Aldrich

Benzyl chloride

Benzyl chloride

ReagentPlus®, 99%, contains ≤1% propylene oxide as stabilizer

Synonym(s):

α-Chlorotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2Cl
CAS Number:
Molecular Weight:
126.58
Beilstein:
471308
MDL number:
UNSPSC Code:
12352300
eCl@ss:
39050202
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor density

4.36 (vs air)

vapor pressure

10.3 mmHg ( 60 °C)
7 mmHg ( 55 °C)

product line

ReagentPlus®

Assay

99%

form

liquid

autoignition temp.

1085 °F

contains

≤1% propylene oxide as stabilizer

expl. lim.

14 %

refractive index

n20/D 1.538 (lit.)

bp

177-181 °C (lit.)

mp

−43 °C (lit.)

solubility

0.46 g/L at 30 °C (Decomposes in contact with water)

density

1.1 g/mL at 25 °C (lit.)

functional group

chloro
phenyl

SMILES string

ClCc1ccccc1

InChI

1S/C7H7Cl/c8-6-7-4-2-1-3-5-7/h1-5H,6H2

InChI key

KCXMKQUNVWSEMD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzyl chloride (BnCl) is an organochlorine compound, which is widely used in organic synthesis for the preparation of useful chemical building blocks.[1][2]

Application

Benzyl chloride can be used as:
  • A benzylating agent for the benzylation of benzene and other aromatic compounds.[1][2]
  • A reactant for the preparation of 4-methoxydiphenylmethane by Friedel-Crafts alkylation with anisole.[3]

It can also be used to introduce the benzyl protecting group to alcohols and carboxylic acids by forming corresponding benzyl ether, and benzyl ester, respectively.[1][4]

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Eye Dam. 1 - Muta. 1B - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Oral - STOT SE 3

Target Organs

Heart,forestomach, Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

152.6 °F - closed cup

Flash Point(C)

67 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Benzylation of benzene by benzyl chloride over iron mesoporous molecular sieves materials
Bachari K, et al.
J. Catal., 221(1), 55-61 (2004)
Benzylation of benzene and other aromatics by benzyl chloride over mesoporous AlSBA-15 catalysts.
Vinu A.,e et al.
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 80(1-3), 195-203 (2005)
Benzylation of toluene by benzyl chloride over protonic zeolites.
Coq B, et al.
Applied Catalysis A: General, 100(1), 69-75 (1993)
Heterogeneous phase alkylation of phenols making use of phase transfer catalysis and microwave irradiation
Keglevich G, et al.
Letters in Organic Chemistry, 6(7), 535-539 (2009)
Design and performance of supported Lewis acid catalysts derived from metal contaminated biomass for Friedel-Crafts alkylation and acylation
Losfeld G, et al.
Catalysis Today, 189(1), 111-116 (2012)

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service