Skip to Content
Merck
All Photos(1)

Key Documents

137030

Sigma-Aldrich

Diethyl 1,4-dihydro-2,4,6-trimethyl-3,5-pyridinedicarboxylate

99%

Synonym(s):

3,5-Dicarbethoxy-1,4-dihydrocollidine, DDC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H21NO4
CAS Number:
Molecular Weight:
267.32
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

99%

form

powder

mp

130-132 °C (lit.)

SMILES string

CCOC(=O)C1=C(C)NC(C)=C(C1C)C(=O)OCC

InChI

1S/C14H21NO4/c1-6-18-13(16)11-8(3)12(14(17)19-7-2)10(5)15-9(11)4/h8,15H,6-7H2,1-5H3

InChI key

CDVAIHNNWWJFJW-UHFFFAOYSA-N

Application

Diethyl 1,4-dihydro-2,4,6-trimethyl-3,5-pyridinedicarboxylate was used to induce Mallory-Denk body formation in mice in vivo[1].

Biochem/physiol Actions

Diethyl 1,4-dihydro-2,4,6-trimethyl-3,5-pyridinedicarboxylate blocks the heme synthesis and prevents the induction of hepatic heme oxygenase-1 in mice[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tohru Itoh et al.
FEBS letters, 583(4), 777-781 (2009-01-29)
In injured livers where hepatocyte growth is severely limited, facultative hepatic stem/progenitor cells, termed oval cells in rodents, are known to emerge and contribute to the regeneration process. Here, we investigated a possible involvement of Wnt signaling during mouse oval
Pavel Strnad et al.
Hepatology (Baltimore, Md.), 48(3), 931-942 (2008-08-13)
The cytoplasmic hepatocyte inclusions, Mallory-Denk bodies (MDBs), are characteristic of several liver disorders, including alcoholic and nonalcoholic steatohepatitis. In mice, MDBs can be induced by long-term feeding with 3,5-diethoxycarbonyl-1,4-dihydrocollidine (DDC) for 3 to 4 months or rapidly reformed in DDC-induced
Volker Mostert et al.
Drug metabolism reviews, 39(2-3), 619-626 (2007-09-06)
Heme oxygenase (HO)-1 is induced by oxidative stress and protects against oxidant injury. We examined the effect of rapid induction of hepatic HO-1 on serum iron level. Serum iron was approximately doubled within 6 h when HO-1 was induced by
Johanna R Schaub et al.
Nature, 557(7704), 247-251 (2018-05-04)
Transdifferentiation is a complete and stable change in cell identity that serves as an alternative to stem-cell-mediated organ regeneration. In adult mammals, findings of transdifferentiation have been limited to the replenishment of cells lost from preexisting structures, in the presence
Leila Jemail et al.
Scientific reports, 8(1), 6415-6415 (2018-04-25)
Kupffer cells (KCs) are key players in maintaining tissue homeostasis and are involved in various liver diseases. However, the roles of KCs in the pathogenesis of cholangiopathy are largely unknown. We aimed to investigate the precise roles of KCs in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service