I3146
ITS Liquid Media Supplement (100×)
liquid, sterile-filtered, BioReagent, suitable for cell culture
Synonym(s):
Human ITS Liquid Media, Insulin-transferrin-sodium selenite
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
sterility
sterile-filtered
product line
BioReagent
form
liquid
technique(s)
cell culture | mammalian: suitable
single cell analysis: suitable
impurities
endotoxin, tested
shipped in
ambient
storage temp.
2-8°C
Application
ITS Liquid Media Supplement (100×) has been used as a media supplement to culture:
- α mouse liver 12 (AML-12) cells
- rat fetal testis explants
- human dental pulp stem cells (DPSC)
Biochem/physiol Actions
- Insulin: a polypeptide hormone that promotes the uptake of glucose and amino acids and may owe an observed mitogenic effect to this property.
- Transferrin: an iron-transport protein.
- Selenium: an essential trace element normally provided by serum.
Components
Contains 1.0 mg/ml recombinant human insulin, 0.55 mg/ml human transferrin (substantially iron-free), and 0.5 μg/ml sodium selenite at the 100x concentration.
Other Notes
ITS is a mixture of recombinant human insulin, human transferrin, and sodium selenite. It is a general cell supplement designed for use in non-complex media (e.g. MEM, RPMI-1640) and complex media (e.g. Ham?s F-12, DME/F-12, MEM) with sodium pyruvate.
Preparation Note
Prepared in Earle′s Balanced Salt Solution (EBSS) without phenol red.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Biomedical materials (Bristol, England), 6(3), 031001-031001 (2011-05-14)
Polyelectrolyte layer-by-layer (LbL) nanofilms are interesting polymeric structures, built by alternating adsorption of positively and negatively charged polyelectrolytes. They consist of multilayer sheets with nanometric overall thickness, and they can be used as supports and surface coatings for in vitro
Veterinary research communications, 36(2), 139-148 (2012-03-07)
In the dog, mesenchymal stem cells (MSCs) have been shown to reside in the bone marrow (bone marrow-derived mesenchymal stem cells: BM-MSCs) as well as in the adipose tissue (adipose tissue-derived stem cells: ADSCs). Potential application fields for these multipotent
Scientific reports, 7(1), 1729-1729 (2017-05-13)
Hepatitis B virus (HBV) infection causes acute hepatitis B (AHB), chronic hepatitis B (CHB), liver cirrhosis (LC), and eventually hepatocellular carcinoma (HCC). The presence of hepatitis B e antigen (HBeAg) in the serum generally indicates ongoing viral replication and disease
Cell death & disease, 9(6), 602-602 (2018-05-24)
Long non-coding RNAs (lncRNAs) are a new class of regulators of various human diseases. This study was designed to explore the potential role of lncRNAs in experimental hepatic damage. In vivo hepatic damage in mice and in vitro hepatocyte damage
PloS one, 9(6), e98063-e98063 (2014-06-04)
MicroRNAs (miRs) play a pivotal role in a variety of biological processes including stem cell differentiation and function. Human foetal femur derived skeletal stem cells (SSCs) display enhanced proliferation and multipotential capacity indicating excellent potential as candidates for tissue engineering
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service