Skip to Content
Merck
All Photos(4)

Documents

U5127

Sigma-Aldrich

Ursodeoxycholic acid

≥99%

Synonym(s):

3α,7β-Dihydroxy-5β-cholan-24-oic acid, 5β-Cholan-24-oic acid-3α,7β-diol, 7β-Hydroxylithocholic acid, UDCS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H40O4
CAS Number:
Molecular Weight:
392.57
Beilstein:
3219888
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

description

anionic detergent

Assay

≥99%

mol wt

392.57 g/mol

mp

203-204 °C (lit.)

SMILES string

C[C@H](CCC(O)=O)[C@H]1CC[C@H]2[C@@H]3[C@@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C

InChI

1S/C24H40O4/c1-14(4-7-21(27)28)17-5-6-18-22-19(9-11-24(17,18)3)23(2)10-8-16(25)12-15(23)13-20(22)26/h14-20,22,25-26H,4-13H2,1-3H3,(H,27,28)/t14-,15+,16-,17-,18+,19+,20+,22+,23+,24-/m1/s1

InChI key

RUDATBOHQWOJDD-UZVSRGJWSA-N

Gene Information

human ... NR1H4(9971)

Looking for similar products? Visit Product Comparison Guide

Application

Ursodeoxycholic acid (UDCS) is a cell protectant used extensively to mitigate hepatic and biliary diseases. Ursodeoxycholic acid may be used to study its specific activities that range from reduction of cholesterol absorpition, cholesterol gallstone dissolution to suppression of immune response.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei Zhou et al.
Journal of biomolecular screening, 15(5), 488-497 (2010-05-08)
Dietary long-chain fatty acid (LCFA) uptake across cell membranes is mediated principally by fatty acid transport proteins (FATPs). Six subtypes of this transporter are differentially expressed throughout the human and rodent body. To facilitate drugs discovery against FATP subtypes, the
Ruchika Sharma et al.
Journal of medicinal chemistry, 54(1), 122-130 (2010-12-17)
Ursodeoxycholic acid (UDCA) is used for the treatment of hepatic inflammatory diseases. Recent studies have shown that UDCA's biological effects are partly glucocorticoid receptor (GR) mediated. UDCA derivatives were synthesized and screened for ability to induce GR translocation in a
Yannick Bacq et al.
Gastroenterology, 143(6), 1492-1501 (2012-08-16)
We performed a meta-analysis to evaluate the effects of ursodeoxycholic acid (UDCA) on pruritus, liver test results, and outcomes of babies born to women with intrahepatic cholestasis of pregnancy (ICP). We performed a systematic review of 9 published, randomized controlled
L Serfaty et al.
Gastroenterologie clinique et biologique, 34(10), 516-522 (2010-07-09)
Colorectal cancer is respectively the third and second most common cancer among men and women in France. Interest in chemoprevention for colorectal cancer has increased over the last two decades. Beside non-steroidal anti-inflammatory drugs, ursodeoxycholic acid (UDCA) may have chemopreventive
Yan Gong et al.
The American journal of gastroenterology, 102(8), 1799-1807 (2007-04-27)
Ursodeoxycholic acid (UDCA) is used for primary biliary cirrhosis (PBC), but the beneficial effects remain controversial. We performed an updated systematic review to evaluate the benefits and harms of UDCA in patients with PBC. We included randomized clinical trials evaluating

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service