Skip to Content
Merck
All Photos(1)

Documents

135615

Sigma-Aldrich

2-Bromo-2-methylpropane

98%

Synonym(s):

2-methyl-2-bromopropane, tert-Butyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CBr
CAS Number:
Molecular Weight:
137.02
Beilstein:
1730892
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

contains

0.5% potassium carbonate as stabilizer

refractive index

n20/D 1.4279 (lit.)

bp

71-73 °C (lit.)

mp

−20 °C (lit.)

solubility

H2O: insoluble
organic solvents: miscible

density

1.22 g/mL at 20 °C (lit.)

SMILES string

CC(C)(C)Br

InChI

1S/C4H9Br/c1-4(2,3)5/h1-3H3

InChI key

RKSOPLXZQNSWAS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Bromo-2-methylpropane (tert-Butyl bromide) is a versatile reactant in organic synthesis, facilitating the introduction of the tert-butyl group. It causes the massive deguanylation of guanine based-nucleosides and massive deadenylation of adenine based-nucleosides.

Application

2-Bromo-2-methylpropane was used to study the massive deadenylation of adenine based-nucleosides induced by halogenated alkanes under physiological conditions.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

60.8 °F - closed cup

Flash Point(C)

16 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deadenylation of Adenine Based-Nucleosides and Calf thymus DNA Induced by Halogenated Alkanes at the Physiological Condition.
Sherchan J, et al.
Bull. Korean Chem. Soc., 30(10), 2318-2328 (2009)
Deguanylation of guanine based-nucleosides and calf thymus DNA induced by halogenated alkanes at the physiological condition.
Sherchan J and Lee E-S.
Bull. Korean Chem. Soc., 30(12), 2318-2328 (2009)
tert-Butyl Bromide-Promoted Intramolecular Cyclization of 2-Arylamino Phenyl Ketones and Its Combination with Cu-Catalyzed C-N Coupling: Synthesis of Acridines at Room Temperature
Z Cao, et al.
The Journal of Organic Chemistry, 85, 10167-10174 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service