Skip to Content
Merck
All Photos(1)

Key Documents

249890

Sigma-Aldrich

1-Bromo-2,2-dimethylpropane

98%

Synonym(s):

Neopentyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CCH2Br
CAS Number:
Molecular Weight:
151.04
Beilstein:
1730989
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.436 (lit.)

bp

105-106 °C/767 mmHg (lit.)

density

1.199 g/mL at 25 °C (lit.)

functional group

alkyl halide
bromo

SMILES string

CC(C)(C)CBr

InChI

1S/C5H11Br/c1-5(2,3)4-6/h4H2,1-3H3

InChI key

CQWYAXCOVZKLHY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

44.6 °F - closed cup

Flash Point(C)

7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carmen E Lisowski et al.
The journal of physical chemistry. A, 114(38), 10395-10402 (2010-09-03)
The recombination of chloromethyl and t-butyl radicals at room temperature was used to generate neopentyl chloride molecules with 89 kcal mol(-1) of internal energy. The observed unimolecular reactions, which give 2-methyl-2-butene and 2-methyl-1-butene plus HCl, as products, are explained by
An improved method for cysteine alkylation.
Perrey DA and Uckun FM.
Tetrahedron Letters, 42(10), 1859-1861 (2001)
J Ashby et al.
Mutation research, 140(2-3), 71-74 (1984-06-01)
Neopentyl bromide and pentaerythrityl tetrachloride were shown here to be non-mutagenic to 7 strains of Salmonella typhimurium. These inactivities are reflected in the inability of either compound to produce a colour reaction in the chemical alkylation test of Epstein (4-nitrobenzylpyridine
Direct mono-N-alkylation of amines in ionic liquids: chemoselectivity and reactivity.
Chiappe C and Pieraccini D.
Green Chemistry, 5(2), 193-197 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service