Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Acetanilide
Synonym(s):
N
-Phenylacetamide, Acetanilide
Linear Formula:
CH
3
CONHC
6
H
5
CAS Number:
103-84-4
Molecular Weight:
135.16
EC Number:
203-150-7
Products
Product Category
building blocks (4)
Brand
Sigma-Aldrich (5)
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Puriss (1)
Puriss P.A. (1)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
112933
Acetanilide,
99%
Expand
View Pricing
8.22344
Acetanilide,
for synthesis
Expand
View Pricing
397229
Acetanilide,
purified by sublimation, ≥99.9%
Expand
View Pricing
397237
Acetanilide,
zone-refined, purified by sublimation, ≥99.95%
Expand
View Pricing
00401
Acetanilide,
puriss. p.a., ≥99.5% (CHN)
Expand
View Pricing