Skip to Content
MilliporeSigma

Skip To

HPA019239

Anti-OSGIN1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Ovary, kidney and liver protein 38, Anti-Oxidative stress-induced growth inhibitor 1, Anti-Pregnancy-induced growth inhibitor OKL38, Anti-huOKL38

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC (p)
Citations:
5


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

immunogen sequence

SGFLTRNQAQQPFSLWARNVVLATGTFDSPARLGIPGEALPFIHHELSALEAATRVGAVTPASDPVLIIGAGLSAADAVLYARHYNIP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... OSGIN1(29948)

General description

The gene OSGIN1 (oxidative stress-induced growth inhibitor 1) is mapped to human chromosome 16q23.3. OSGIN1 transcripts are ubiquitously expressed in all tissues with strong expression in liver, kidney, ovary and testis. The protein localizes in the nucleus and mitochondria.[1] OSGIN1 is commonly referred as OKL38 (ovary, kidney and liver protein 38).

Immunogen

Oxidative stress-induced growth inhibitor 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

OSGIN1 (oxidative stress-induced growth inhibitor 1) is down-regulated in kidney tumors, breast cancer cells. Increase in OSGIN1 levels causes growth inhibition and cell death. Upon DNA damage, OSGIN1 works together with p53 and regulates mitochondria function. It induces cytochrome c response and mediates apoptosis.[1]

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Other Notes

Corresponding Antigen APREST74494

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Tara A McCannel et al.
Investigative ophthalmology & visual science, 52(6), 3018-3022 (2010-08-07)
To identify genomic targets for ciliochoroidal melanoma diagnosis, prognosis, and therapy. Fifty-eight ciliochoroidal melanomas were analyzed by high-resolution, genome-wide, single nucleotide polymorphism (SNP) mapping arrays. The 58 SNP arrays were compared to 48 HapMap normals representing both sexes and assessed
H Huynh et al.
Endocrinology, 142(8), 3607-3615 (2001-07-19)
Growth factors and growth inhibitors play crucial roles in the growth regulation and differentiation of mammary epithelial cells. Studies have shown that during pregnancy, with the onset of terminal differentiation, there is a dramatic decrease in the proliferation of the
Choon Kiat Ong et al.
The Journal of biological chemistry, 279(1), 743-754 (2003-10-23)
We previously demonstrated the growth inhibitory property of OKL38 and its possible roles in mammary carcinogenesis. To further understand the regulation and roles of OKL38 in tumorigenesis we proceeded to clone and characterize the human OKL38 gene and three of



Global Trade Item Number

SKUGTIN
HPA019239-100UL04061836312732

Questions

Reviews

No rating value

Active Filters