Skip to Content
MilliporeSigma

Skip To

HPA010554

Anti-TPBG antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-5T4 oncofetal antigen, Anti-5T4 oncofetal trophoblast glycoprotein, Anti-5T4 oncotrophoblast glycoprotein, Anti-M6P1, Anti-Trophoblast glycoprotein precursor

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC
Citations:
4


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL, immunohistochemistry: 1:50-1:200

immunogen sequence

LVSLTYVSFRNLTHLESLHLEDNALKVLHNGTLAELQGLPHIRVFLDNNPWVCDCHMADMVTWLKETEVVQGKDRLTCAYPEKMRNRVLLELNSADLDCDPILP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TPBG(7162)

General description

TPBG (trophoblast glycoprotein) is an oncofetal glycoprotein with a molecular weight of 72kDa. It is rarely expressed in normal adult tissues, but it is seen to be expressed in multiple cancerous tissues. It is also expressed by trophoblast cells and placenta. It is a cell surface protein. Its extracellular domain is highly glycosylated and contains eight leucine-rich repeats (LRRs). This protein is composed of 420 amino acids and contains a single transmembrane domain.[1]

Immunogen

Trophoblast glycoprotein precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

TPBG (trophoblast glycoprotein) interacts with human trophoblast. This protein is highly expressed in pancreatic ductal adenocarcinoma (PDAC), which is linked with tumor-node-metastasis (TNM) stage of the tumor. It is also involved in metastasis in PDAC.[1] This protein promotes EMT (epithelial-mesenchymal transition) and CXCL12/CXCR4 (C-X-C motif chemokine ligand/CXC receptor) chemotaxis, and inhibits canonical Wnt/β-catenin. It is overexpressed in malignant pleural mesothelioma (MPM), and has potential as an immunotherapy target in the same.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Other Notes

Corresponding Antigen APREST71498

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

or

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Ping He et al.
Molecular medicine reports, 12(1), 503-509 (2015-03-05)
Trophoblast glycoprotein (TPBG), a 72 kDa glycoprotein was identified using a monoclonal antibody, which specifically binds human trophoblast. The expression of TPBG in normal tissues is limited; however, it is upregulated in numerous types of cancer. When TPBG is expressed
Yuguang Zhao et al.
Structure (London, England : 1993), 22(4), 612-620 (2014-03-04)
The tumor antigen 5T4/WAIF1 (Wnt-activated inhibitory factor 1; also known as Trophoblast glycoprotein TPBG) is a cell surface protein targeted in multiple cancer immunotherapy clinical trials. Recently, it has been shown that 5T4/WAIF1 inhibits Wnt/β-catenin signaling, a signaling system central to
Peter L Stern et al.
Seminars in cancer biology, 29, 13-20 (2014-07-30)
Oncofoetal antigens are present during foetal development with generally limited expression in the adult but are upregulated in cancer. These molecules can sometimes be used to diagnose or follow treatment of tumours or as a target for different immunotherapies. The



Global Trade Item Number

SKUGTIN
HPA010554-100UL04061837125270

Questions

Reviews

No rating value

Active Filters