Skip to Content
MilliporeSigma

Skip To

HPA009035

Anti-GALNT5 antibody produced in rabbit

Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-GalNAc-T5

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
100 μL
Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)
$667.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Human Protein Atlas Number:
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IF, IHC
Citations:
3

$667.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL, immunohistochemistry: 1:200-1:500

immunogen sequence

MKPPLRGHGKGAWGKENVRKTEESVLKVEVDLDQTQRERKMQNALGRGKVVPLWHPAHLQTLPVTPNKQKTDGRGTKPEASSHQGTPKQTTAQGAPKTSFIAAKGTQVVKISVHMGRVSLKQEPR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GALNT5(11227)

General description

GALNT5 (N-acetylgalactosaminyltransferase 5) is a highly tissue specific protein. It predominantly localizes to the cytoplasm of gastric epithelial and gastric cancer cells.[1] This gene localizes to human chromosome 2q24.

Immunogen

polypeptide N-acetylgalactosaminyltransferase 5

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

GALNT5 (N-acetylgalactosaminyltransferase 5) glycosylates restricted types of substrates. It is capable of interacting with Golgi proteins, by projecting its catalytic domain in the lumen of the Golgi complex. It might play a role in maintaining the structure of gastric mucosa, and less differentiated gastric tumor tissues show low expression of this protein. Its low expression is linked with TNM classification, and functions as an independent prognostic marker for poor outcome in gastric cancer patients.[1] In breast cancer, missense mutation in this gene results in the loss of enzymatic function of this GALNT5 protein. This gene is also up-regulated in hepatoblastomas when compared to the expression in differentiated livers.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Other Notes

Corresponding Antigen APREST71963

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
HPA008963HPA011140HPA012628
Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

product line

Prestige Antibodies® Powered by Atlas Antibodies

product line

Prestige Antibodies® Powered by Atlas Antibodies

product line

Prestige Antibodies® Powered by Atlas Antibodies

product line

Prestige Antibodies® Powered by Atlas Antibodies

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice


Still not finding the right product?

or

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Muhammad Ramzan Manwar Hussain et al.
Journal of cellular biochemistry, 115(2), 313-327 (2013-09-17)
Aberrant glycosylation by N-acetylgalactosaminyl transferases (GALNTs) is a well-described pathological alteration that is widespread in hereditary diseases, prominently including human cancers, familial tumoral calcinosis and hyperostosis-hyperphosphatemia. In this study, we integrated different computational tools to perform the in silico analysis
Tatiane Cristina Rodrigues et al.
Future oncology (London, England), 10(15), 2449-2457 (2014-12-20)
Cytogenetic data of hepatoblastomas, a rare embryonal tumor of the liver, mostly consist of descriptions of whole-chromosome aneuploidies and large chromosome alterations. High-resolution cytogenetics may provide clues to hepatoblastoma tumorigenesis and indicate markers with clinical significance. We used array-CGH (180K)
H He et al.
British journal of cancer, 110(8), 2021-2029 (2014-03-13)
The family of polypeptide N-acetylgalactosaminyl transferases is responsible for the altered O-linked glycosylation occurring during the development of various cancers and their progression via altering O-glycan biosynthesis. Our studies were designed to investigate the expression and prognostic values of GalNAc-T5



Global Trade Item Number

SKUGTIN
HPA009035-25UL04061842797660
HPA009035-100UL04061836300258

Questions

Reviews

No rating value

Active Filters