Skip to Content
MilliporeSigma

Skip To

HPA005701

Anti-PRELID1 antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-25 kDa protein of relevant evolutionary and lymphoid interest antibody produced in rabbit, Anti-PRELI antibody produced in rabbit, Anti-Px19-like protein antibody produced in rabbit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
100 μL
Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)
$667.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
Human Protein Atlas Number:
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IF, IHC
Citations:
3

$667.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL, immunohistochemistry: 1:50-1:200

immunogen sequence

VAHSVYVLEDSIVDPQNQTMTTFTWNINHARLMVVEERCVYCVNSDNSGWTEIRREAWVSSSLFGVSRAVQEFGLARFKSNVTKTMKGFEYILAKLQGEAPSKTLVETAKEAKEKAKETALAATEKA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PRELID1(27166)

General description

The gene encoding PRELI domain-containing protein 1 (PRELID1) is present on chromosome 5q34-q35. The protein′s subcellular location is the mitochondrion.

Immunogen

Px19-like protein recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

PRELI domain-containing protein 1 (PRELID1) is a factor influencing the human primary T-helper (Th)-cell death and differentiation. It induces oxidative stress and a mitochondrial apoptosis pathway in the primary Th cells. PRELID1 also inhibits Th2-cell development by down-regulating a signal transducer and activator of transcription 6 (STAT6), a key transcription factor involved in Th2 differentiation. By associating with dynamin-like GTPase Optic Atrophy-1 (OPA1), it helps in mitochondrial morphology maintenance. PRELID1 upholds mitochondrial membrane potential and enhances respiratory chain (RC) function, by inducing complex-I/NADH dehydrogenase and ATP synthase expression.[1]

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Other Notes

Corresponding Antigen APREST86540

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
HPA003600HPA005558HPA000848
Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

technique(s)

immunofluorescence: 0.25-2 μg/mL, immunohistochemistry: 1:50-1:200

technique(s)

immunoblotting: 0.04-0.4 μg/mL, immunofluorescence: 0.25-2 μg/mL, immunohistochemistry: 1:200-1:500

technique(s)

immunofluorescence: 0.25-2 μg/mL, immunohistochemistry: 1:200-1:500

technique(s)

immunohistochemistry: 1:50- 1:200

UniProt accession no.

Q9Y255

UniProt accession no.

Q15691

UniProt accession no.

Q14145

UniProt accession no.

Q9UBX5


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

Prestige Antibodies Immunofluorescence Procedure


Johanna Tahvanainen et al.
Blood, 113(6), 1268-1277 (2008-10-24)
The identification of novel factors regulating human T helper (Th)-cell differentiation into functionally distinct Th1 and Th2 subsets is important for understanding the mechanisms behind human autoimmune and allergic diseases. We have identified a protein of relevant evolutionary and lymphoid
M R McKeller et al.
Cell death & disease, 1, e21-e21 (2011-03-03)
Proteins containing the late embryogenesis abundant (LEA) motif comprise a conserved family, postulated to act as cell protectors. However, their function and mechanisms of action remain unclear. Here we show that PRELI, a mammalian LEA-containing homolog of yeast Ups1p, can
Elizabeth J Fox et al.
The Biochemical journal, 378(Pt 3), 817-825 (2003-12-03)
The characterization of mitochondrial proteins is important for the understanding of both normal cellular function and mitochondrial disease. In the present study we identify a novel mitochondrial protein, PRELI (protein of relevant evolutionary and lymphoid interest), that is encoded within



Global Trade Item Number

SKUGTIN
HPA005701-25UL04061842769629
HPA005701-100UL04061835679713

Questions

Reviews

No rating value

Active Filters