Skip to Content
MilliporeSigma

Skip To

HPA005543

Anti-IFI6 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Ifi-6-16, Anti-Interferon alpha-inducible protein 6 precursor, Anti-Interferon-induced protein 6-16

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

immunogen sequence

AANSVAASLMSWSAILNGGGVPAGGLVATLQSLGAGGSSVVIGNIGALMGYATHKYLDSEEDE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IFI6(2537)

General description

IFI6 (interferon, α-inducible protein 6) is a part of ISG12 (IFN stimulated gene) family. In humans, there are four members in this family, where IFI6 is located at chromosome 1p35, and the other three at 14q32. It is a hydrophobic protein with a molecular weight of 12.9kDa. It has a conserved ISG12 motif, which is made of ~80 amino acids.[1] When expressed ectopically, IFI6 is found to be localized to mitochondria.[2]

Immunogen

Interferon alpha-inducible protein 6 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The functional characteristics of IFI6 (interferon, α-inducible protein 6) is not yet well known.[2] This protein is induced by interferons (IFN), and is responsible for cell survival.[3] It is strongly induced by IFN-α2b, where it prevents apoptosis and maintains mitochondrial stability. This way, it also antagonizes the apoptotic effect of TRAIL, especially in tumor cells. Thus, it can be a potential therapeutic target for various malignancies, such as myeloma.[2] IFI6 is also induced by estrogen, and is highly expressed in the initial stages of breast cancer. It alters the balance between pro- and anti-apoptotic members of Bcl-2 family. Hence, it plays a role in poor prognosis of breast cancer, hyperplasia and tamoxifen resistance.[3]

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Other Notes

Corresponding Antigen APREST86654.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



V Cheriyath et al.
Oncogene, 31(17), 2222-2236 (2011-10-15)
Hormonally regulated survival factors can have an important role in breast cancer. Here we elucidate G1P3, a survival protein induced by interferons (IFNs), as a target of estrogen signaling and a contributor to poor outcomes in estrogen receptor-positive (ER(+)) breast
Venugopalan Cheriyath et al.
The Journal of clinical investigation, 117(10), 3107-3117 (2007-09-08)
The effectiveness of IFN-alpha2b for human multiple myeloma has been variable. TRAIL has been proposed to mediate IFN-alpha2b apoptosis in myeloma. In this study we assessed the effects of IFN-alpha2b signaling on the apoptotic activity of TRAIL and human myeloma
Nadeene Parker et al.
BMC genomics, 5(1), 8-8 (2004-01-20)
The human 6-16 and ISG12 genes are transcriptionally upregulated in a variety of cell types in response to type I interferon (IFN). The predicted products of these genes are small (12.9 and 11.5 kDa respectively), hydrophobic proteins that share 36%