36144
MCPA methyl ester
PESTANAL®, analytical standard
Synonym(s):
Methyl (4-chloro-2-methylphenoxy)acetate
About This Item
Recommended Products
grade
analytical standard
Quality Level
product line
PESTANAL®
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
NMR: suitable
gas chromatography (GC): suitable
refractive index
n20/D 1.52-1.53
suitability
passes test for identity (NMR)
application(s)
agriculture
environmental
format
neat
SMILES string
COC(=O)COc1ccc(Cl)cc1C
InChI
1S/C10H11ClO3/c1-7-5-8(11)3-4-9(7)14-6-10(12)13-2/h3-5H,6H2,1-2H3
InChI key
VWERIRLJUWTNDA-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Recommended products
Legal Information
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1
Storage Class
11 - Combustible Solids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service