Skip to Content
MilliporeSigma

Skip To

8.00413

Ethyl chloroacetate

for synthesis

Synonym(s):

Ethyl chloroacetate, Chloroacetic acid ethyl ester

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C4H7ClO2
CAS Number:
Molecular Weight:
122.55
EC Index Number:
203-294-0
UNSPSC Code:
12352108
MDL number:


vapor pressure

4.8 hPa ( 20 °C)

Quality Level

assay

≥99.0% (GC)

autoignition temp.

452 °C

potency

180 mg/kg LD50, oral (Rat), 161 mg/kg LD50, skin (Rat)

expl. lim.

2.6 % (v/v)

bp

144 °C/1013 hPa

mp

-26 °C

transition temp

flash point 54 °C

solubility

12.3 g/L

density

1.15 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C4H7ClO2/c1-2-7-4(6)3-5/h2-3H2,1H3

InChI key

VEUUMBGHMNQHGO-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.149 - 1.151
Identity (IR): passes test
Evaporation Number: 0.23


Still not finding the right product?

Explore all of our products under Ethyl chloroacetate


signalword

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class

3 - Flammable liquids

wgk

WGK 3

flash_point_f

129.2 °F - closed cup

flash_point_c

54 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library