Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W393304

Sigma-Aldrich

cis-3-Hexenyl propionate

≥97%, stabilized, FG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H16O2
CAS Number:
Molecular Weight:
156.22
FEMA Number:
3933
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.564
NACRES:
NA.21
organoleptic:
apple; green; leafy; fruity; pear
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 872/2012

assay

≥97%

contains

α-tocopherol as stabilizer (synthetic)

refractive index

n20/D 1.43 (lit.)

bp

83 °C/17 mmHg (lit.)

density

0.887 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

apple; green; leafy; fruity; pear

SMILES string

[H]\C(CC)=C(/[H])CCOC(=O)CC

InChI

1S/C9H16O2/c1-3-5-6-7-8-11-9(10)4-2/h5-6H,3-4,7-8H2,1-2H3/b6-5-

InChI key

LGTLDEUQCOJGFP-WAYWQWQTSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

150.0 °F - closed cup

flash_point_c

65.56 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Angel Llopis-Giménez et al.
Journal of chemical ecology, 46(11-12), 1017-1031 (2020-11-06)
Insect chemosensation is crucial for many aspects related to food seeking, enemy avoidance, and reproduction. Different families of receptors and binding proteins interact with chemical stimuli, including odorant receptors (ORs), ionotropic receptors (IRs), gustatory receptors (GRs), odorant binding proteins (OBPs)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service