T6008
Transaldolase from baker′s yeast (S. cerevisiae)
lyophilized powder, 10-30 units/mg protein (biuret)
Synonym(s):
D-Sedoheptulose-7-phosphate: D-Glyceraldehyde-3-phosphate dihydroxyacetonetransferase
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
lyophilized powder
specific activity
10-30 units/mg protein (biuret)
storage temp.
−20°C
Application
Useful in systems requiring low sulfate concentrations.
Unit Definition
One unit will produce 1.0 μmole of D-glyceraldehyde 3-phosphate from D-fructose 6-phosphate per min in the presence of D-erythrose 4-phosphate, at pH 7.7 at 25°C in a coupled system with GDH/TPI and β-NADH.
Physical form
Lyophilized, essentially sulfate-free; contains approx. 5% citrate buffer salts
signalword
Danger
hcodes
pcodes
Hazard Classifications
Resp. Sens. 1
Storage Class
11 - Combustible Solids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Anne K Samland et al.
Chembiochem : a European journal of chemical biology, 12(10), 1454-1474 (2011-05-17)
Aldol reactions constitute a powerful methodology for carbon-carbon bond formation in synthetic organic chemistry. Biocatalytic carboligation by aldolases offers a green, uniquely regio- and stereoselective tool with which to perform these transformations. Recent advances in the field, fueled by both
Andras Perl et al.
Trends in molecular medicine, 17(7), 395-403 (2011-03-08)
Metabolism of glucose through the pentose phosphate pathway (PPP) influences the development of diverse pathologies. Hemolytic anemia due to deficiency of PPP enzyme glucose 6-phosphate dehydrogenase is the most common genetic disease in humans. Recently, inactivation of another PPP enzyme
Anja Lehwess-Litzmann et al.
Nature chemical biology, 7(10), 678-684 (2011-08-23)
We examined the catalytic cycle of transaldolase (TAL) from Thermoplasma acidophilum by cryocrystallography and were able to structurally characterize--for the first time, to our knowledge--different genuine TAL reaction intermediates. These include the Schiff base adducts formed between the catalytic lysine
Irene González-Rodríguez et al.
Applied and environmental microbiology, 78(11), 3992-3998 (2012-03-27)
The ability of bifidobacteria to establish in the intestine of mammals is among the main factors considered to be important for achieving probiotic effects. The role of surface molecules from Bifidobacterium bifidum taxon in mucin adhesion capability and the aggregation
Cun Wang et al.
Cancer letters, 313(2), 154-166 (2011-10-26)
Hepatocellular carcinoma (HCC) is one of serious disorders with the highest morbidities and mortalities worldwide. Metastasis is the major concern that causes death in HCC. The goal of this study was to screen and identify potential serum proteins indicating HCC
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service