Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML2462

Sigma-Aldrich

HZ-166

≥98% (HPLC)

Synonym(s):

8-Ethynyl-6-(2-pyridinyl)-4H-Imidazo[1,5-a][1,4]benzodiazepine-3-carboxylic acid ethyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$55.75
25 MG
$245.00

$55.75

List Price$60.40Save 8%
Web-Only Promotion

Available to ship onMay 05, 2025Details


Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
5 MG
$55.75
25 MG
$245.00

About This Item

Empirical Formula (Hill Notation):
C21H16N4O2
CAS Number:
Molecular Weight:
356.38
UNSPSC Code:
12352200
NACRES:
NA.77

$55.75

List Price$60.40Save 8%
Web-Only Promotion

Available to ship onMay 05, 2025Details


Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

C#CC1=CC=C2C(C(C3=NC=CC=C3)=NCC4=C(C(OCC)=O)N=CN42)=C1

InChI key

ZSEDMNMFODMQRD-UHFFFAOYSA-N

Biochem/physiol Actions

HZ-166 is a potent, non-sedative GABAA receptor subtype-selective benzodiazepine site agonist exhibits preference toward α2- and α3-GABAARs. HZ-166 exhibits antihyperalgesic properties in mice models of neuropathic and inflammatory pain, without sedation and motor impairment.
potent, non-sedative GABAA receptor subtype-selective benzodiazepine site agonist exhibits preference toward α2- and α3-GABAARs

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service