Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1412430

Sigma-Aldrich

ANTI-MUC2 antibody produced in mouse

clone 4A4, purified immunoglobulin, buffered aqueous solution

Synonym(s):

MLP, MUC2, SMUC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4A4, monoclonal

form

buffered aqueous solution

mol wt

antigen 36.63 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MUC2(4583)

General description

Mucin 2, oligomeric mucus/gel-forming (MUC2) is encoded by the gene mapped within 400kb on human chromosome 11p15.5. The encoded protein has a molecular mass of ~550 kDa and is expressed at high levels in the intestine and at lower levels in the respiratory tree. Mucin is a key component of mucus and it consists of one partial von Willebrand domain (vWD) at N-terminal and two complete domains including CysD domain and two proline, threonine and serine (PTS) domains that become densely O-glycosylated to form the prolonged mucin domains.
This gene encodes a member of the mucin protein family. Mucins are high molecular weight glycoproteins produced by many epithelial tissues. The protein encoded by this gene is secreted and forms an insoluble mucous barrier that protects the gut lumen. The protein polymerizes into a gel of which 80% is composed of oligosaccharide side chains by weight. The protein features a central domain containing tandem repeats rich in threonine and proline that varies between 50 and 115 copies in different individuals. Alternatively spliced transcript variants of this gene have been described, but their full-length nature is not known. (provided by RefSeq)

Immunogen

MUC2 (NP_002448, 5081 a.a. ~ 5179 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
TTEVSYAGCTKTVLMNHCSGSCGTFVMYSAKAQALDHSCSCCKEEKTSQREVVLSCPNGGSLTHTYTHIESCQCQDTVCGLPTGTSRRARRSPRHLGSG*

Biochem/physiol Actions

Mucin 2, oligomeric mucus/gel-forming (MUC2) acts as a major constituent of the two-layered mucous structure in the colon. MUC2 in the outer mucous layer provides the habitat for the commensal flora and in the stratified inner mucous layer it protects the epithelial cells from bacteria invasion. The encoded protein controls expression and antimicrobial activity of β-defensin 2. Deficiency of MUC2 mucin might lead to opportunistic microbial invasion and/or impaired innate responses.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E R Cobo et al.
Mucosal immunology, 8(6), 1360-1372 (2015-04-30)
In this study we identified mechanisms at the colonic mucosa by which MUC2 mucin regulated the production of β-defensin in a proinflammatory milieu but functionally protected susceptible bacteria from its antimicrobial effects. The regulator role of MUC2 on production of
L E Vinall et al.
Human genetics, 102(3), 357-366 (1998-04-17)
A family of four genes that encode major secreted mucins (MUC6, MUC2, MUC5AC and MUC5B) map to within 400 kb on chromosome 11p15.5. These genes contain long stretches of tandem repeats of sequence that encode serine- and threonine-rich domains but
P Pigny et al.
Genomics, 38(3), 340-352 (1996-12-15)
Four distinct genes that encode mucins have previously been mapped to chromosome 11p15.5. Three of these genes (MUC2, MUC5AC, and MUC6) show a high level of genetically determined polymorphism and were analyzed in the CEPH families. Linkage analysis placed all
Christian V Recktenwald et al.
The Journal of biological chemistry, 291(26), 13580-13590 (2016-04-30)
The main structural component of the mucus in the gastrointestinal tract is the MUC2 mucin. It forms large networks that in colon build the loose outer mucous layer that provides the habitat for the commensal flora and the inner mucous
Sjoerd van der Post et al.
Journal of proteome research, 13(12), 6013-6023 (2014-11-19)
The polymeric mucin MUC2 constitutes the main structural component of the mucus that covers the colon epithelium. The protein's central mucin domain is highly O-glycosylated and binds water to provide lubrication and prevent dehydration, binds bacteria, and separates the bacteria

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service