Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

77661

Sigma-Aldrich

FabRICATOR®

for cleaving 5 mg IgG, 5000 U/vial

Synonym(s):

IdeS from Streptococcus pyogenes

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
3.4.22
UNSPSC Code:
12352204

recombinant

expressed in E. coli

form

powder

specific activity

5000 U/vial

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Unit Definition

One unit is defined as the amount of enzyme required to fragment 95% of 1 ug of human IgG in 30 minutes at 37°C, pH 6.6 as monitored by SDS-PAGE.

Analysis Note

SDS gel electrophoresis ≥ 95 % purity

Legal Information

FabRICATOR is a registered trademark of Genovis AB

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sialylation-independent mechanism involved in the amelioration of murine immune thrombocytopenia using intravenous gammaglobulin.
Leontyev, D., et al.
Transfusion, doi: 10-doi: 10 (2012)
Johnson Agniswamy et al.
The Journal of biological chemistry, 279(50), 52789-52796 (2004-10-07)
Group A Streptococcus has evolved numerous mechanisms to evade the host immune system to survive, disseminate, and cause disease. Recently a secreted protein named Mac-1 was identified and shown to enhance survival of the pathogen. A new variant of Mac-1
Mary H Ryan et al.
Molecular immunology, 45(7), 1837-1846 (2007-12-26)
A comparative in vitro survey of physiologically relevant human and microbial proteinases defined a number of enzymes that induced specific hinge domain cleavage in human IgG1. Several of these proteinases have been associated with tumor growth, inflammation, and infection. A
Theo Rispens et al.
Journal of the American Chemical Society, 133(26), 10302-10311 (2011-06-02)
Immunoglobulin G (IgG) antibodies are symmetrical molecules that may be regarded as covalent dimers of 2 half-molecules, each consisting of a light chain and a heavy chain. Human IgG4 is an unusually dynamic antibody, with half-molecule exchange ("Fab-arm exchange") resulting
M Lišková et al.
Analytical and bioanalytical chemistry, 400(2), 369-379 (2011-02-08)
A number of biologically important molecules, such as DNA, proteins, and antibodies, are routinely conjugated with fluorescent tags for high-sensitivity analyses. Here, the application of quantum dots in the place of bright and size-tunable luminophores is studied. Several selected bioconjugation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service