Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

51804

Sigma-Aldrich

TRIS pH 8.0; 0.05 M − Magnesium chloride 10 mM − Cobalt(III)-hexamine 1.0 mM − Ethanol 20% solution

Synonym(s):

Crystallization solution 13/Kit-No 80701

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

pH

8.0±0.2 ( in neat)

density

0.98 g/mL at 20 °C

storage temp.

2-8°C

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

95.0 °F

flash_point_c

35 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service